Skip to Content
Merck
All Photos(1)

Key Documents

MABT140

Sigma-Aldrich

Anti-Claudin-4 Antibody, clone 10B4.3

clone 10B4.3, from mouse

Synonym(s):

Claudin-4, Clostridium perfringens enterotoxin receptor, CPE-R, CPE-receptor, Williams-Beuren syndrome chromosomal region 8 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€358.00

€358.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€358.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€358.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

10B4.3, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CLDN4(1364)

General description

Claudin 4, also known as Clostridium perfringens enterotoxin receptor, or CPE-R, or CPE-receptor or Williams-Beuren syndrome chromosomal region 8 protein, and encoded by the gene CLDN4/CPER/CPETR1/WBSCR8, plays a critical role in tight junction specific obliteration of intercellular space. Claudin 4 directly interacts with several important tight junction associated proteins including ZO-1,2,3 and EPHA2 proteins, and through these interactions, Claudin 4 regulates tight junction formation. Claudin 4 is localized to cell junctions and membranes at tight junctions. Claudin 4 is expressed in most tissues particularly in intestine, liver, and pancreas. Defects in Claudin 4 may be associated with Williams Beuren syndrome genetic deletion syndrome. Claudin 4 is also over expressed in many cancers and thus is a target of anti-cancer research using C. perfringens enterotoxin.

Immunogen

Epitope: Cytoplasmic domain
KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human Claudin-4.

Application

Anti-Claudin-4 Antibody, clone 10B4.3 is an antibody against Claudin-4 for use in western blotting & IHC (Paraffin).
Research Category
Cell Structure
Research Sub Category
ECM Proteins
Western Blotting Analysis: 2.0 µg/mL from a representative lot detected Claudin-4 in 10 µg of MCF-7 cell lysate.

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Claudin-4 in human ileum and human tonsil tissue.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Claudin-4 in 10 µg of HeLa cell lysate.

Target description

~ 22 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service