Skip to Content
Merck
All Photos(1)

Key Documents

MABT121

Sigma-Aldrich

Anti-Vimentin Antibody, clone 3CB2

clone 3CB2, from mouse

Synonym(s):

Vimentin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€487.00

€487.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€487.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€487.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3CB2, monoclonal

species reactivity

chameleon (GLIA 15:389-400 (1995), Prada, F. A., et. al.), rat (GLIA 15:389-400 (1995), Prada, F. A., et. al.), human, lizard

species reactivity (predicted by homology)

chicken (immunogen homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VIM(7431)

General description

Vimentin is a class III intermediate filament protein (homopolymers) found in various non-epithelial cells including mesenchymal cells and fibroblasts. In many cells of mesenchymal origin, it is one of the most prominent phosphoproteins, especially during cell division when enhanced phosphorylation of vimentin corresponds with the reorganization of vimentin filaments.

Immunogen

Vimentin extracted from chicken embryonic brain tissue.

Application

Anti-Vimentin Antibody, clone 3CB2 is an antibody against Vimentin for use in WB, IC, IH.
Immunocytochemistry Analysis: A representative lot was used by an independent laboratory in human central and peripheral retinal sections (Perez-Alvarez, M.J., et al. (2008). J Neurosci Res. 86(8):1871-1883).

Western Blot Analysis:
A representative lot was used to detect vimentin in chicken and rat total brain extracts (Prada, FA., et al. (1995). GLIA 15:389-400).

Immunohistochemistry Analysis:
A representative lot was used to detect vimentin in E3 chick embryo sections and different regions of the developing chick central nervous system (Prada, FA., et al. (1995). GLIA 15:389-400).
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Western Blot in ARPE19 cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Vimentin in ARPE19 cell lysate.

Target description

~54 kDa observed

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
ARPE cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Qi et al.
Experimental and therapeutic medicine, 15(4), 3509-3515 (2018-03-17)
The present study aimed to investigate the effects of JWA knockout (JWA-/-) on malignant transformation of murine embryonic fibroblast (MEF) cells using a conditional JWA-/- mouse model. Once MEF cells were prepared, the potential role of JWA-/- on proliferation, migration
Yi Sui et al.
British journal of cancer, 125(7), 1003-1015 (2021-07-12)
Metastasis is the major cause of treatment failure and cancer-related deaths in prostate cancer (PCa) patients. Our previous study demonstrated that a CD44+ subpopulation isolated from PCa cells or tumours possesses both stem cell properties and metastatic potential, serving as
Yi Sui et al.
Annals of translational medicine, 9(1), 45-45 (2021-02-09)
Prostate cancer (PCa) is one of the most common types of cancer and the emerging resistance to androgen deprivation therapy in PCa aggravates disease progression. In this study, we examined the potential pro-tumorigenic functions of NANOGP8 in prostate cancer development.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service