Skip to Content
Merck

Skip To

MABS522

Anti-PIKFYVE Antibody, clone R159.4.3C9

clone R159.4.3C9, from mouse

Synonym(s):

1-phosphatidylinositol 3-phosphate 5-kinase, Epididymis luminal protein 37, FYVE finger-containing phosphoinositide kinase, PIKfyve, Phosphatidylinositol 3-phosphate 5-kinase type III, PIPkin-III, Type III PIP kinase, Zinc finger FYVE domain containing 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
R159.4.3C9, monoclonal
Application:
WB
Citations:
2
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

R159.4.3C9, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PIKFYVE(200576)

General description

1-phosphatidylinositol 3-phosphate 5-kinase (EC 2.7.1.150; UniProt Q9Y2I7; also known as Epididymis luminal protein 37, FYVE finger-containing phosphoinositide kinase, PIKfyve, Phosphatidylinositol 3-phosphate 5-kinase type III, PIPkin-III, Type III PIP kinase, Zinc finger FYVE domain containing 29) is encoded by the PIKFYVE (also known as CFD, FAB1, HEL37, KIAA0981, PIP5K, PIP5K3, ZFYVE29) gene (Gene ID 200576) in human. PIKfyve is a class III lipid kinase that is responsible for phosphorylating phosphatidylinositol (PtdIns) and phosphatidylinositol 3-phosphate (PtdIns3P) of their inositol ring at the D-5 position to form PtdIns3P and PtdIns(3,5)P2 (phosphatidylinositol 3,5-bisphosphate), respectively. PIKfyve forms a stable ternary heterooligomeric complex with its regulator ArPIKfyve (encoded by the VAC14 gene) and the Sac1 domain-containing PtdIns(3,5)P2 5-phosphatase Sac3 (encoded by the FIG4 gene), and both ArPIKfyve and Sac3 are required for maximal PIKfyve activity. PIKfyve is also known to interact with the Rab9 effector RABEPK and the kinesin adaptor JLP (encoded by SPAG9). By regulating endosomal trafficking, PIKfyve plays an important role in multiple biological processes, such as GLUT4 translocation, retroviral budding, autophagy, and neurodegeneration. Human PIKfyve is initially produced as a 2098-amino acid protein whose N-terminal initiator methionine is removed posttranslationally. PIKFYVE gene mutation is linked to corneal fleck dystrophy (CFD) characterized by white flecks scattered in all levels of the stroma.
~240/50 kDa observed. 237,1 kDa (isoform 1), 50.21 kDa (isoform 2), 51.48 kDa (isoform 3), 61.60 kDa (isoform 4) calculated.

Immunogen

AviTag-6xHis-tagged recombinant protein corresponding an internal fragment of human PIKFYVE.
Epitope: Internal (N-terminal half).

Application

Anti-PIKFYVE Antibody, clone R159.4.3C9 is an antibody against PIKFYVE for use in Western Blotting.
Research Category
Signaling
Research Sub Category
General Post-translation Modification
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected PIKFYVE in 10 µg of HepG2 cell lysate.

Biochem/physiol Actions

Expected to react with all four spliced isoforms of human PIKfyve reported by UniProt (Q9Y2I7-1 through Q9Y2I7-4).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HL-60 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected PIKFYVE in 20 µg of HL-60 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4500758MABC195SAB1300387
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

mouse

biological source

rabbit

biological source

mouse

biological source

rabbit

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

IgG fraction of antiserum

clone

R159.4.3C9, monoclonal

clone

polyclonal

clone

7F11.1, monoclonal

clone

polyclonal

species reactivity

human

species reactivity

mouse, human

species reactivity

human

species reactivity

mouse, human

Gene Information

human ... PIKFYVE(200576)

Gene Information

human ... PIKFYVE(200576)

Gene Information

human ... ZFYVE26(23503)

Gene Information

human ... PIP5K2A(1023434)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters