Skip to Content
Merck

MABS481

Anti-PL Scramblase 3, clone 3B4 Antibody

clone 3B4, from mouse

Synonym(s):

Phospholipid scramblase 3, PL scramblase 3, Ca(2+)-dependent phospholipid scramblase 3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
3B4, monoclonal
Application:
WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3B4, monoclonal

species reactivity

human, mouse

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PLSCR3(57048)

General description

Phospholipid scramblase 3 (PL scramblase 3), also known as Ca(2+)-dependent phospholipid scramblase 3, and encoded by the gene name PLSCR3, is a member of the phospholipid scramblase family. Recent studies have indicated that PL scramblase 3 may mediate accelerated ATP-independent bidirectional transbilayer migration of phospholipids upon binding calcium ions that results in a loss of phospholipid asymmetry in the plasma membrane. PL scramblase 3 is suggested to play a central role in the initiation of fibrin clot formation, in the activation of mast cells and in the recognition of apoptotic and injured cells by the reticuloendothelial system. It may also play a role in apoptosis, through translocation of cardiolipin from the inner to the outer mitochondrial membrane which promotes BID recruitment and enhances tBid-induced mitochondrial damages. PL scramblase 3 has been demonstrated to interact with PDCD6 in a calcium-dependent manner.
~31 kDa observed

Immunogen

Recombinant protein corresponding to human PL Scramlase 3.

Application

This Anti-PL Scramblase 3 antibody is validated for use in Western Blotting for the detection of PL Scramblase 3.
Western Blotting Analysis: A representative lot detected PL Scramlase 3 in SH-SY5Y cell lysate (Chu, C.T., et al. (2013). Nature Cell Biology. 15(10):1197-1205).
Western Blotting Analysis: A representative lot detected PL Scramlase 3 in primary adipocytes lysate (Wiedmer, T., et al. (2004). PNAS. 101(36):13296-13301).

Physical form

Format: Purified

Analysis Note

Evaluated by Western Blotting in NIH-3T3/L1 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected PL Scramlase 3 in 10 µg of NIH-3T3/L1 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library