Skip to Content
Merck

Skip To

MABS461

Anti-Neutrophil Elastase Antibody, clone AHN-10

clone AHN-10, from mouse

Synonym(s):

Neutrophil elastase, Bone marrow serine protease, Elastase-2, Human leukocyte elastase, HLE, Medullasin, PMN elastase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 416,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
AHN-10, monoclonal
Application:
ICC, IHC, IP, RIA, inhibition assay
Citations:
2

€ 416,00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1927

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AHN-10, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, inhibition assay: suitable, radioimmunoassay: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ELANE(1991)

General description

28 kDa calculated
Neutrophil Elastase, also known as Bone marrow serine protease, Elastase-2, Human leukocyte elastase (HLE), Medullasin or PMN elastase, and encoded by the gene ELANE/ELA2, is a serine proteinase in the same family as chymotrypsin. Neutrophil Elastase has broad substrate specificity and is part of our host defense mechanism against bacteria. Unfortunately passively released or actively secreted elastase from neutrophils has been linked to the pathologic processes of a variety of inflammatory diseases, including idiopathic pulmonary fibrosis, rheumatoid arthritis, adult respiratory distress syndrome, and cystic fibrosis. Neutrophil Elastase is localized to the cytoplasm in granules. Neutrophil Elastase is expressed in neutrophils and macrophages. Defects in Neutrophil Elastase may be associated with cyclic neutropenia and congential neutropenia diseases where neutrophils fail to mature leaving the patient very vunderable to infections.

Immunogen

Whole cells corresponding to human Neutrophil Elastase.

Application

Detect Elastase using this mouse monoclonal antibody, Anti-Neutrophil Elastase Antibody, clone AHN-10 validated for use in ICC, IHC, IP, Radioimmunoassay & Inhibition.
Immunohistochemistry Analysis: A representative lot from an independent laboratory detected Neutrophil Elastase in unirradiated and irradiated human and mouse skin tissues (Rhodes, L. E., et al. (2009). FASEB J. 23(11): 3947-3956.; Nicolaou, A., et al. (2012). Photochem Photobiol Sci. 11(2):371-380.; Rutter, J. J., et al. (2009). 129(12):2861-2867.).

Western Blotting Analysis: A representative lot from an independent laboratory detected Neutrophil Elastase in human pancreatic tissue lysate (Mayerle, J., et al. (2005). Gastroenterology. 129(4):1251-1267.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated Neutrophil Elastase in human pancreatic tissue lysate (Mayerle, J., et al. (2005). Gastroenterology. 129(4):1251-1267.).

Immunocytochemistry Analysis: A representative lot from an independent laboratory detected Neutrophil Elastase in human acinar cells (Mayerle, J., et al. (2005). Gastroenterology. 129(4):1251-1267.).

Inhibition Analysis: This assay inhibits the elastinolytic actiity of purified human Neutrophil Elastase (Skubitz, K., et al. (1988). J Leukoc Biol. 44(3):158-165.).

Radioimmunoassay Analysis: A representative lot from an independent laboratory detected Neutrophil Elastase using purified Neutrophil Elastase (Skubitz, K., et al. (1988). J Leukoc Biol. 44(3):158-165.).
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Biochem/physiol Actions

This antibody detects with human Neutrophil Elastase but not other major neutrophil proteases: Cathapsin G, proteinase 3, collagenase, or esterase N (Skubitz, K. M., et al. (1988). J Leukoc Biol. 44(3):158-165.).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Immunocytochemistry Analysis in peripheral blood mononuclear cells.

Immunocytochemistry Analysis: A 1:250 dilution of this antibody detected Neutrophil Elastase in peripheral blood mononuclear cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB5500151SAB1305716481001
species reactivity

human

species reactivity

human (tested)

species reactivity

human

species reactivity

human

Gene Information

human ... ELANE(1991)

Gene Information

human ... ELANE(1991)

Gene Information

human ... ELANE(1991)

Gene Information

human ... ELANE(1991)

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified antibody

clone

AHN-10, monoclonal

clone

SP203, monoclonal

clone

polyclonal

clone

polyclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

rabbit


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters