Skip to Content
Merck

Skip To

MABS460

Anti-FGF23 Antibody, clone 278-2

clone 278-2, from mouse

Synonym(s):

Fibroblast growth factor 23, FGF-23, Phosphatonin, Tumor-derived hypophosphatemia-inducing factor, Fibroblast growth factor 23 N-terminal peptide, Fibroblast growth factor 23 C-terminal peptide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 402,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
278-2, monoclonal
Application:
FACS, WB
Citations:
-

€ 402,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

278-2, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FGF23(8074)

General description

Fibroblast growth factor 23 (FGF-23) is also called Phosphatonin and Tumor-derived hypophosphatemia-inducing factor. FGF-23 belongs to the heparin-binding growth factors family and has several important functions including: regulation of phosphate homeostasis, inhibition of renal tubular phosphate transport, upregulation of EGR1 expression, regulation of vitamin-D metabolism, and negative regulation of osteoblast differentiation and matrix mineralization. FGF-23 is expressed in osteogenic cells, adult trabecular bone, osteocytes and flattened bone-lining cells (inactive osteoblasts). Defects in FGF23 can lead to autosomal dominant hypophosphataemic rickets (ADHR) and hyperphosphatemic familial tumoral calcinosis (HFTC).
~25 kDa observed

Immunogen

Corresponding to human FGF23.

Application

Anti-FGF23 Antibody, clone 278-2 is a mouse monoclonal antibody & validated for use in western blotting.
Flow Cytometry Analysis: 2 µg from a representative lot detected FGF23 in Jurkat and human PMBC cells.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG1κ in buffer containing PBS with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human salivary gland tissue lysate
Evaluated by Western Blotting in human salivary gland tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected FGF23 in 10 µg of human salivary gland tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4503210ABN442SAB2502007
Gene Information

human ... FGF23(8074)

Gene Information

human ... FGF23(8074)

Gene Information

human ... FGF20(26281)

Gene Information

mouse ... Fgf23(64654)

species reactivity

human

species reactivity

rat, human, mouse

species reactivity

human

species reactivity

rat, mouse

clone

278-2, monoclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

goat

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody


Still not finding the right product?

Explore all of our products under Anti-FGF23 Antibody, clone 278-2

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters