Skip to Content
Merck
All Photos(3)

Key Documents

MABS453

Sigma-Aldrich

Anti-Axin2, clone cond4-1D5 Antibody

clone cond4-1D5, from rat

Synonym(s):

Axin-2, Axin-like protein, Axil, Conductin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€367.00

€367.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€367.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€367.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

cond4-1D5, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AXIN2(8313)

General description

Axin-2, or Axis inhibition protein 2, is an inhibitor of the Wnt signaling pathway that down-regulates beta-catenin and APC through phosphorylation. Axin-2 abnormalities are linked to defective mismatch repair and have been found to be involved in colorectal cancer (CRC) and oligodontia-colorectal cancer syndrome (ODCRCS). Axin-2 can be found in brain and lymphoblast tissues.

Specificity

This monoclonal antibody recognizes both endogeneous and overexpressed Axin2 and does not cross-react with Axin1

Immunogen

Linear peptide corresponding to Human Axin 2.

Application

Immunocytochemistry Analysis: 20 μg/mL of this antibody from a representative lot detected Axin2 in HeLa, HEK293 and NIH-3T3 cells.
Western Blotting Analysis: A representative lot of this antibody detected Axin2 in SW 480 cell lines
Research Category
Signaling

Apoptosis & Cancer
Research Sub Category
General Post-translation Modification
This Anti-Axin2, clone cond4-1D5 Antibody is validated for use in Western Blotting and Immunocytochemistry for the detection of Axin2.

Quality

Evaluated by Western Blotting in HCT116 cell lysate.

Western Blotting Analysis: 1 μg/mL of this antibody detected Axin2 in 10 µg of HCT116 cell lysate.

Target description

~ 95 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service