Skip to Content
Merck
All Photos(1)

Key Documents

MABS2310

Sigma-Aldrich

Anti-TROP2 Antibody, clone TrMab-29

Synonym(s):

Tumor-associated calcium signal transducer 2;Cell surface glycoprotein Trop-2;Membrane component chromosome 1 surface marker 1;Pancreatic carcinoma marker protein GA733-1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€142.00
100 μG
€357.00

€142.00


Estimated to ship on29 March 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€142.00
100 μG
€357.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
clone:
TrMab-29, monoclonal
application:
FACS
IHC (p)
WB
species reactivity:
human
technique(s):
flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
citations:

€142.00


Estimated to ship on29 March 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

TrMab-29, monoclonal

mol wt

calculated mol wt 35.71 kDa
observed mol wt ~38 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... DAG1(1605)

General description

Tumor-associated calcium signal transducer 2 (UniProt: P09758; also known as Cell surface glycoprotein Trop-2, Trophoblast antigen 2, Membrane component chromosome 1 surface marker 1, Pancreatic carcinoma marker protein GA733-1) is encoded by the TACSTD2 (also known as GA733-1, M1S1, TROP2) gene (Gene ID: 4070) in human. TROP2 is a single-pass type 1 membrane glycoprotein that is synthesized with a signal peptide (aa 1-26), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 27-274), a transmembrane domain (aa 275-297), and a cytoplasmic domain (aa 298-323). It contains both extracellular epidermal growth factor-like and thyroglobulin type-1 repeat domains. Its expression is observed in placenta and is highly expressed in many cancers where it plays a role in tumor progression. About 85% of all solid cancers display increased expression of TROP2. Its expression is also reported in stem cells in various tissues. TROP2 expression in cancer cells has been correlated with drug resistance. Upregulation of TROP2 is reported to drive the expression and activation of CREB1, Jun, NF-kB, Rb, STAT1, and STAT3 through induction of the cyclin D1 and the ERK pathways. Mutations in TACSTD2 is known to cause corneal dystrophy of the gelatinous drop-like type (GDLD), an autosomal recessive disorder that is characterized by pathognomonic branching lattice figures in the cornea leading to blindness. (Ref.: Sayama, Y., et al. (2021). Biochem. Biophys. Rep. 25; 100902; Anna Shvartsur, A., and Bonavida, B. (2015). Genes Cancer. 6(3-4); 84-105).

Specificity

Clone TrMab-29 is a mouse monoclonal antibody that detects Tumor-associated calcium signal transducer 2 TROP2).

Immunogen

CHO K1 cells overexpressing TROP2-PA and PA-TROP2-RAP-MAP.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from CHO-K1 cells overexpressing TROP2.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected TROP2 in lysate from CHO-K1 cells overexpressing TROP2

Tested Applications


Immunohistochemistry (Paraffin) Analysis: A representative lot detected TROP2 in Immunohistochemistry applications (Sayama, Y., et al. (2021). Biochem Biophys Rep. 25: 100902).

Western Blotting Analysis: A representative lot detected TROP2 in Western Blotting applications (Sayama, Y., et al. (2021). Biochem Biophys Rep. 25: 100902).

Flow Cytometry Analysis: A representative lot detected TROP2 in Flow Cytometry applications (Sayama, Y., et al. (2021). Biochem Biophys Rep. 25: 100902).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-TROP2, clone TrMab-29, Cat. No. MABS2310, is a mouse monoclonal antibody that detects TROP2 and is tested for use in Flow Cytometry, Immunohistochemistry, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service