Skip to Content
Merck
All Photos(2)

Key Documents

MABS2238

Sigma-Aldrich

Anti-ELOVL5 Antibody, clone Z88

Synonym(s):

Elongation of very long chain fatty acids protein 5;EC:2.3.1.199;3-keto acyl-CoA synthase ELOVL5;ELOVL fatty acid elongase 5;ELOVL FA elongase 5;Fatty acid elongase 1;hELO1;Very long chain 3-ketoacyl-CoA synthase 5;Very long chain 3-oxoacyl-CoA synthase 5

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€140.00
100 μG
€353.00

€140.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€140.00
100 μG
€353.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€140.00


Estimated to ship on26 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

Z88, monoclonal

mol wt

calculated mol wt 35.29 kDa
observed mol wt ~38 kDa

purified by

using protein G

species reactivity

human

species reactivity (predicted by homology)

monkey, bovine

packaging

antibody small pack of 100 μg

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ELOVL5(60481)

General description

Elongation of very long chain fatty acids protein 5 (UniProt: EC:2.3.1.199, 3-keto acyl-CoA synthase ELOVL5, ELOVL fatty acid elongase 5, ELOVL FA elongase 5, Fatty acid elongase 1, hELO1, Very long chain 3-ketoacyl-CoA synthase 5, Very long chain 3-oxoacyl-CoA synthase 5) is encoded by the ELOVL5 (also known as EVOVL2, PRO0530) gene (Gene ID: 60481) in human. ELOVL5 is a widely expressed, multi-pass membrane protein found in the endoplasmic reticulum (ER). Higher expression of ELOVL5 is reported in the adrenal gland and testis. It catalyzes the first and rate-limiting reaction of the four reactions that constitute the long-chain fatty acids elongation cycle. It exhibits marked preference for polyunsaturated molecules. It allows the addition of 2 carbons to the chain of long- and very long-chain fatty acids (VLCFAs) per cycle. Significantly higher levels of ELOVL5 are reported in prostate cancer compared to nonmalignant prostate. ELOVL5 depletion is reported to markedly alter mitochondrial morphology and function that leads to excess generation of reactive oxygen species and results in suppression of prostate cancer cell proliferation, in vivo tumor growth, and metastasis. ELOVL5 is expressed in the brain in a region- and cell type-specific manner and is abundant in the cerebellum, brainstem, and primary and accessory olfactory regions. Mutations of ELOVL5 gene are known to cause the spino-cerebellar ataxia type 38 (SCA38), a rare autosomal neurological disease characterized by gait abnormality, dysarthria, dysphagia, hyposmia, and peripheral neuropathy. (Ref.: Balbo, I., et al. (2021). Front. Neuroanat. 15; 669073; Centenera, MM., et al. (2021). Cancer Res. 81(7); 1704-1718; Alnabulsi, A., et al. (2019). Int. J. Cancer. 145(4);1138-1147).

Specificity

Clone Z88 is a mouse monoclonal antibody that detects Elongation of very long chain fatty acids protein 5 (ELOVL5). It targets an epitope within 9 amino acids from the C-terminal region.

Immunogen

Ovalbumin-conjugated linear peptide corresponding to 9 amino acids from the C-terminal region of human Elongation of very long chain fatty acids protein 5 (ELOVL5).

Application

Quality Control Testing

Evaluated by Western Blotting in MCF-7 cell lysates.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ELOVL5 in MCF-7 cell lysates.

Tested Applications

Western Blotting Analysis: A representative lot detected ELOVL5 in Western Blotting applications (Alnabulsi, A., et al. (2019). Int J Cancer. 145(4):1138-1147).

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected ELOVL5 in human prostate tissue sections.

ELISA Analysis: A representative lot detected ELOVL5 in ELISA applications (Alnabulsi, A., et al. (2019). Int J Cancer. 145(4):1138-1147).

Immunohistochemistry Applications: A representative lot detected ELOVL5 in Immunohistochemistry applications (Alnabulsi, A., et al. (2019). Int J Cancer. 145(4):1138-1147).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-ELOVL5 clone Z88, Cat. No. MABS2238, is a mouse monoclonal antibody that detects ELOVL5 and is tested for use in ELISA, Immunohistochemistry, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service