Skip to Content
Merck
All Photos(1)

Key Documents

MABS1950

Sigma-Aldrich

Anti-p15 Antibody, clone PAT56B

clone PAT56B, from rat

Synonym(s):

Cyclin-dependent kinase 4 inhibitor B, p14-INK4b, p15-INK4b

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€116.00
100 μG
€353.00

€116.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
€116.00
100 μG
€353.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€116.00


Check Cart for Availability

Request a Bulk Order

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PAT56B, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Cdkn2B(12579)

General description

Cyclin-dependent kinase 4 inhibitor B (UniProt: P55271; also known as p14-INK4b, p15-INK4b) is encoded by the Cdkn2b gene (Gene ID: 12579) in murine species. p15-INK4b is a ubiquitously expressed member of the INK4 family of cyclin-dependent kinase inhibitors. It induces a G1-phase cell cycle arrest through heterodimerization with cyclin-dependent kinase 4/6, thereby serves as a tumor suppressor. It also serves as a potential effector of TGF-beta induced cell cycle arrest. p15 also plays an important role in the regulation of cellular commitment of hematopoietic progenitor cells and myeloid cell differentiation. It contains four ankyrin repeats (aa 5-34; 38-66; 71-100; and 104-130). p15 shares extensive homology with p16 and serves an important backup function for p16 in senescence. Cdkn2b gene is frequently hypermethylated in myeloid neoplasia. Genes coding for both p15 and p16 are reported to often deleted in human tumors. p15 can serve as a good prognostic marker for monitoring the response to treatment with DNA methylation inhibitors. (Ref.: Wolff, L., and Bies, J. (2013). Blood Cells Mol. Dis. 50(4); 227-231).

Specificity

Clone PAT56B is a rat monoclonal antibody that specifically detects murine Cyclin-dependent kinase 4 inhibitor B (p15-INK4b).

Immunogen

GST/His-tagged recombinant full-length mouse Cyclin-dependent kinase 4 inhibitor B (p15-INK4b).

Application

Anti-p15, clone PAT56B, Cat. No. MABS1950, is a rat monoclonal antibody that detects Cyclin-dependent kinase 4 inhibitor B (p15-INK4b) and has been tested for use in Immunohistochemistry and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected p15 in Immunohistochemistry applications (Munoz-Espin, D., et. al. (2013). Cell. 155(5):1104-18).
Research Category
Signaling

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysates.

Western Blotting Analysis: 4 µg/mL of this antibody detected p15 in NIH/3T3 cell lysates.

Target description

~14 kDa observed; 13.79 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service