Skip to Content
Merck
All Photos(1)

Key Documents

MABS1929

Sigma-Aldrich

Anti-hLYVE-1 Antibody, clone 8C

clone 8C, from mouse

Synonym(s):

Lymphatic vessel endothelial hyaluronic acid receptor 1, Cell surface retention sequence-binding protein 1, CRSBP-1, Extracellular link domain-containing protein 1, Hyaluronic acid receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8C, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... LYVE1(10894)

General description

Lymphatic vessel endothelial hyaluronic acid receptor 1 (UniProt: Q9Y5Y7; LYVE-1, Cell surface retention sequence-binding protein 1, CRSBP-1,Extracellular link domain-containing protein 1,Hyaluronic acid receptor) is encoded by the LYVE1 (also known as CRSBP1, HAR, XLKD1,UNQ230/PRO263) gene (Gene ID: 10894) in human. LYVE-1 is a disulfide-linked, homodimeric, single-pass type I membrane protein that undergoes ligand-dependent internalization and recycling at the cell surface. It serves as a ligand-specific transporter trafficking between intracellular organelles (TGN) and the plasma membrane. It is mainly expressed in endothelial cells lining lymphatic vessels. It is expressed on both the luminal and basolateral surfaces of lymphatic endothelium. It is reported to mediate the uptake of intermediate and high molecular weight hyaluronan (HA) and serves as a low affinity receptor for HA-induced lymphoangiogenesis. It also controls the entry and/or migration of leukocytes in lymphatic vessels. The capacity of LYVE-1 to bind HA is based on a receptor distribution and organization of the glycosaminoglycans. (Ref.: Lawrence, W et al. (2016). J Biol Chem. 291(15): 8014 8030).

Specificity

Clone 8C detects Lymphatic vessel endothelial hyaluronic acid receptor 1. It targets an epitope within the ectodomain region.

Immunogen

A recombinant fragment corresponding to the ectodomain (1-232) region of human LYVE-1.

Application

Anti-hLYVE-1, clone 8C, Cat. No. MABS1929, is a mouse monoclonal antibody that detects Lymphatic vessel endothelial hyaluronic acid receptor 1 (LYVE1) and has been tested for use in Flow Cytometry, Immunocytochemistry, and Western Blotting.
Immunocytochemistry Analysis: A representative lot detected hLYVE-1 in Immunocytochemistry applications (Lawrance, W., et. al. (2016). J Biol Chem. hLYVE-1; Banerji, S., et. al. (1999). J Cell Biol. 144(4):789-801).

Western Blotting Analysis: A representative lot detected hLYVE-1 in Western Blotting applications (Banerji, S., et. al. (1999). J Cell Biol. 144(4):789-801; Banerji, S., et. al. (2016). J Biol Chem. 291(48):25004-25018).

Flow Cytometry Analysis: A representative lot detected hLYVE-1 in Flow Cytometry applications (Lawrance, W., et. al. (2016). J Biol Chem. hLYVE-1; Banerji, S., et. al. (2016). J Biol Chem. 291(48):25004-25018).

Quality

Evaluated by Western Blotting in lysates from wild-type and LYVE-1 transfected HEK293T cells.

Western Blotting Analysis: 2 ug/mL of this antibody detected LYVE1 (Lymphatic vessel endothelial hyaluronic acid receptor 1) in lysate from wild-type and LYVE transfected HEK293T cells.

Target description

~60 kDa observed; 35.21 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service