Skip to Content
Merck
All Photos(1)

Key Documents

MABS1913

Sigma-Aldrich

Anti-TAZ Antibody, clone 11

clone 11, from mouse

Synonym(s):

WW domain-containing transcription regulator protein 1, Transcriptional coactivator with PDZ-binding motif

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€131.00
100 μG
€343.00

€131.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
€131.00
100 μG
€343.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€131.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

11, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Wwtr1(97064)

General description

WW domain-containing transcription regulator protein 1 (UniProt: Q9EPK5; also known as Transcriptional coactivator with PDZ-binding motif, TAZ) is encoded by the Wwtr1 (also known as Taz) gene (Gene ID: 97064) in murine species. TAZ is an oncogenic transcriptional activator that acts as a downstream regulatory target in the Hippo signaling pathway that plays a pivotal role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. TAZ is highly expressed in kidney, heart, placenta, and lung tissues. Two isoforms of TAZ have been reported that are produced by alternative splicing. TAZ is concentrated along specific portions of the plasma membrane and also accumulates in punctate nuclear bodies. The ability of TAZ to regulate gene transcription is very tightly regulated through the phosphorylation-dependent binding of 14-3-3 protein. Upon phosphorylation TAZ is retained in cytoplasm by 14-3-3 zeta. Expression of NPHP9 is reported to inhibit the interaction of wild-type TAZ with 14-3-3 and NPHP9 is reported to abolish the phosphorylation of TAZ at Serine 89. TAZ is shown to be involved in non-small-cell lung cancer and its higher expression levels are observed in breast cancer and in breast cancer stem cells. It also regulates embryonic stem-cell self-renewal and promotes cell proliferation and epithelial-mesenchymal transition. TAZ binds to transcription factors via its WW domain, which is localized to amino acids 124-157. Its PZD-binding motif is located in amino acids 389-395 and this motif is essential for stimulated gene transcription.

Specificity

Clone 11 specifically detects WW domain-containing transcription regulator protein 1 (TAZ) in murine species.

Immunogen

His-tagged Recombinant Protein corresponding to Mouse WW domain-containing transcription regulator protein 1 (TAZ).

Application

Anti-TAZ, clone 11, Cat. No. MABS1913, is a mouse monoclonal antibody that detects WW domain-containing transcription regulator protein 1 (TAZ) and has been tested for use in ELISA, Immunoprecipitation, and Western Blotting.
Western Blotting Analysis: 1 ug/mL from a representative lot detected TAZ in mouse lung tissue lysate.

Immunoprecipitation Analysis: A representative lot immunoprecipitated TAZ in Immunoprecipitation applications (Habbig, S., et. al. (2012). Hum Mol Genet. 21(26):5528-38).

Western Blotting Analysis: A representative lot detected TAZ in Western Blotting applications (Habbig, S., et. al. (2012). Hum Mol Genet. 21(26):5528-38).

ELISA Analysis: A representative lot detected TAZ in ELISA applications (Habbig, S., et. al. (2012). Hum Mol Genet. 21(26):5528-38).

Quality

Evaluated by Western Blotting in mouse kidney tissue lysate.

Western Blotting Analysis: 1 ug/mL of this antibody detected TAZ in mouse kidney tissue lysate.

Target description

~55 kDa observed; 43.62 and 49.83 kDa for isoform 1 and 2, respectively. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service