Skip to Content
Merck
All Photos(3)

Key Documents

MABS1911

Sigma-Aldrich

Anti-Serpin B9 Antibody, clone 7D8

clone 7D8, from mouse

Synonym(s):

Cytoplasmic antiproteinase 3, CAP-3, CAP3, Peptidase inhibitor 9, PI-9

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€131.00
100 μL
€337.00

€131.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
€131.00
100 μL
€337.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€131.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7D8, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... SERPINB9(5272)

General description

Serpin B9 (UniProt: P50453; also known as Cytoplasmic antiproteinase 3, CAP-3, CAP3, Peptidase inhibitor 9, PI-9) is encoded by the SERPINB9 (also known as PI9) gene (Gene ID: 5272) in human. Serpins serve as intracellular or extracellular regulators of a wide range of physiological processes such as complement activation, blood coagulation and apoptosis. Within the vertebrate immune system, they control proteases of the classical and innate complement systems, and are also potent inhibitors of many leukocyte granule proteases. Serpin B9 has a broad tissue distribution, being present at high and relatively stable levels natural killer cells and CD8+ T cells, antigen-presenting cells, in many endothelial and mesothelial cells and at sites of immune privilege such as testis and placenta. Serpin B9 serves as a regulator granzyme B activity by acting as an inhibitor of its activity. Overexpression of Serpin B9 may prevent cytotoxic T-lymphocytes from eliminating certain tumor cells. Certain tumors may upregulate Serpin B9 levels as a mechanism of immune evasion. (Ref.: Kaiserman, D., and Bird, PI (2010). Cell Death Differen. 17(4); 586-595).

Specificity

Clone 7D8 detects Serpin B9 in human cells.

Immunogen

His-tagged full length human recombinant Serpin B9.

Application

Anti-Serpin B9, clone 7D8, Cat. No. MABS1911, is a mouse monoclonal antibody that detects Serpin B9 and has been tested for use in ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, and Western Blotting.
ELISA Analysis: A representative lot detected Serpin B9 in ELISA applications (Hirst, C.E., et. al. (2001). Mol Hum Peprod. 7(12):1133-42).

Flow Cytometry Analysis: A representative lot detected Serpin B9 in Flow Cytometry applications (Hirst, C.E., et. al. (2003). J Immunol. 170(2):805-15).

Immunocytochemistry Analysis: A representative lot detected Serpin B9 in Immunocytochemistry applications (Hirst, C.E., et. al. (2003). J Immunol. 170(2):805-15).

Immunofluorescence Analysis: A representative lot detected Serpin B9 in Immunofluorescence applications (Hirst, C.E., et. al. (2003). J Immunol. 170(2):805-15).

Immunohistochemistry Analysis: A representative lot detected Serpin B9 in Immunohistochemistry applications (Hirst, C.E., et. al. (2001). Mol Hum Peprod. 7(12):1133-42).

Western Blotting Analysis: A 1:100 dilution from a representative lot detected Serpin B9 in YT and Jurkat cell lysates (Courtesy of Prof. Phillip Bird, Ph.D., Monash University, Victoria Australia).

Western Blotting Analysis: A representative lot detected Serpin B9 in Western Blotting applications (Hirst, C.E., et. al. (2001). Mol Hum Peprod. 7(12):1133-42; Hirst, C.E., et. al. (2003). J Immunol. 170(2):805-15; Mangan, M.S., et. al. (2015). J Biol Chem. 291(7):3626-38).

Immunocytochemistry Analysis: A representative lot detected Serpin B9 in COS-1 cells (Courtesy of Prof. Phillip Bird, Ph.D., Monash University, Victoria Australia).
Research Category
Inflammation & Immunology

Quality

Evaluated by Immunohistochemistry in human spleen tissue sections.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected Serpin B9 in human spleen tissue sections.

Target description

42.40 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service