Skip to Content
Merck
All Photos(2)

Key Documents

MABS1709

Sigma-Aldrich

Anti-R-Spondin 2 Antibody, clone 14C7.1

clone 14C7.1, from mouse

Synonym(s):

Roof plate-specific spondin-2, hRspo2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€154.00

€154.00


Estimated to ship on13 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€154.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€154.00


Estimated to ship on13 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

14C7.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RSPO2(340419)

General description

R-spondin-2 (UniProt: Q6UXX9; also known as Roof plate-specific spondin-2, hRspo2) is encoded by the RSPO2 (also known as UNQ9384/PRO34209) gene (Gene ID: 340419) in human. R-spondin-2 is a secreted protein that acts as an activator of the canonical Wnt signaling pathway by acting as a ligand for LGR receptors. Upon binding to LGR receptors (LGR4, LGR5 or LGR6) these receptors associate with phosphorylated LRP6 and frizzled receptors, which are activated by extracellular Wnt receptors. These events trigger the canonical Wnt signaling pathway to increase expression of target genes. R-spondin-2 also regulates the canonical Wnt/beta-catenin-dependent pathway and non-canonical Wnt signaling pathway by acting as an inhibitor of ZNRF3, an E3 ubiquitin-protein ligase that serves as an important regulator of the Wnt signaling pathway. In humans, R-Spondin 2 is expressed in organs of endodermal origin in adults, including intestine and lungs and is reported to downregulated in tumors involving these organs. In the embryonic mice it is abundantly expressed in the hippocampus and in developing bones.

Specificity

Clone 14C7.1 specifically detects human R-spondin-2. It targets an epitope within 74 amino acids from the C-terminal end.

Immunogen

GST/His-tagged recombinant fragment corresponding to 74 amino acids from the C-terminal end of human R-Spondin 2.

Application

Anti-R-Spondin 2, clone 14C7.1, Cat. No. MABS1709, is a mouse monoclonal antibody that specifically targets R-Spondin 2 (RSPO2) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution of this antibody detected RSPO2 in human pancreas, human kidney, and human bone marrow tissue.
Research Category
Signaling

Quality

Evaluated by Western Blotting in human recombinant R-spondin 2 protein.

Western Blotting Analysis: 0.02 µg/mL of this antibody detected 0.05 µg of human recombinant R-spondin 2 protein.

Target description

~35 kDa observed; 28.31 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service