Skip to Content
Merck
All Photos(2)

Key Documents

MABS1476

Sigma-Aldrich

Anti-SGK1 Antibody, clone 1G2.1

clone 1G2.1, from mouse

Synonym(s):

Serine/threonine-protein kinase Sgk1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1G2.1, monoclonal

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SGK1(6446)

General description

Serine/threonine-protein kinase Sgk1 (EC 2.7.11.1; UniProt O00141; also known as Serum/glucocorticoid-regulated kinase 1) is encoded by the SGK1 (also known as SGK) gene (Gene ID 6446) in human. SGK1 is a serine/threonine kinase that is induced in response to a variety of stimuli, including serum, osmotic shock, FSH, glucocorticoids, and mineralocorticoids. It is expressed in most tissues with highest levels observed in in the pancreas, followed by placenta, kidney, and lung. SGK1 is involved in the regulation of a wide variety of ion channels, membrane transporters, cellular enzymes, transcription factors, neuronal excitability, cell growth, proliferation, survival, migration and apoptosis. It is also reported to play an important role in cellular stress response. It is also involved in the regulation of renal sodium retention and potassium elimination. SGK1 activity is regulated by phosphorylation and is activated by phosphorylation on Ser422 by mTORC2. This phosphorylation transforms it into a substrate for PDPK1, which phosphorylates it on Thr256. Phosphorylation on Ser397 and Ser401 are also considered to be essential for its activity. It is negatively regulated by ubiquitination and proteasome degradation. High levels of expression of SGK1 gene may contribute to conditions such as hypertension and diabetic nephropathy. SGK1 has been reported to be up-regulated in non-small cell lung cancer (NSCLC) and its over-expression promotes the growth and migration of NSCLC cells via GSK-3 beta phosphorylation and activation of beta-catenin/TCF signaling.


Ref.: Xiaobo, Y et al. (2016) Gene 576, 339-346.

Specificity

Clone 1G2.1 was raised against SGK1 N-terminal fragment, reactivity towards human spliced isoforms 2-5 and murine spliced isoforms 2 and 3 are possible only if the epitope lies outside the first 25-amino acids of isoform 1.

Immunogen

GST-tagged recombinant human SGK1 fragment corresponding to the region N-terminal to the kinase domain.

Application

Detect SGK1 using this mouse monoclonal Anti-SGK1 Antibody, clone 1G2.1, Cat. No. MABS1476, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected SGK1 immunoreactivity in human pancreas, mouse cerebellum, and rat uterus tissue sections.

Western Blotting Analysis: 1 µg/mL from a representative lot detected SGK1 in 10 µg of human (A431, HEK293, HeLa, HepG2, HUVEC, Jurkat, PC3), mouse (NIH/3T3, Raw 264.7), and rat (L6, PC12) cell lysates, as well as in 10 µg of human placenta and mouse brain tissue homogenates.
Research Category
Signaling

Quality

Evaluated by Western Blotting in mouse C2C12 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected SGK1 in 10 µg of mouse C2C12 cell lysate.


Target description

~55 kDa observed. 48.94/48.93/48.93 kDa (human/mouse/rat isoform 1) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riki Ogasawara et al.
The Journal of physiology, 598(23), 5453-5466 (2020-09-08)
Muscle contractions increase protein synthesis in a mechanistic target of rapamycin (mTOR)-dependent manner, yet it is unclear which/how mTOR complexes regulate muscle protein synthesis. We investigated the requirement of mTOR Complex 2 (mTORC2) in contraction-stimulated muscle protein synthesis. mTORC2 inhibition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service