Skip to Content
Merck
All Photos(1)

Key Documents

MABS1219

Sigma-Aldrich

Anti- LTP/CETP Antibody, clone 3-11D

clone 3-11D, from mouse

Synonym(s):

Cholesteryl ester transfer protein, CETP, Lipid transfer protein I, LTP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3-11D, monoclonal

species reactivity

human, mouse, rabbit

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CETP(1071)

General description

Cholesteryl ester transfer protein (UniProt P22687; also known as CETP, Lipid transfer protein I, LTP) is encoded by the CETP (also known as HDLCQ10) gene (Gene ID 1071) in Oryctolagus cuniculus (Rabbit) species. Cholesteryl ester transfer protein (CETP) or lipid transfer protein (LTP) mediates the transport/exhanges of triglycerides and cholesteryl esters (CEs) between very-low-density lipoproteins (VLDL)/low-density lipoproteins (LDL) and high-density lipoproteins (HDL) in both directions. CETP plays an important role in the reverse cholesterol transport (RCT) pathway that mediates the transport of cholesterol from peripheral cells and tissues to the liver for metabolism and excretion in the bile. CETP shuttles CEs from HDL particles to apolipoprotein B-/apoB-containing particles in exchange for triglycerides. CETP is thought to facilitate the generation of small, dense LDL (sd LDL) particles through an indirect mechanism of increased rate of triglyceride transfer from VLDL in exchange for CEs in LDL and HDL. CE transfer between lipoproteins plays a crucial role in intravascular LDL remodeling. Decreased levels of HDL cholesterol and elevated levels of LDL is a risk factor for atherosclerosis and coronary heart disease (CHD).

Specificity

Clone 3-11D (a.k.a. 3-9F) reacts with non-reduced, but not reduced, LTP/CETP (Ko, K.W., et al. (1994). J. Biol. Chem. 269(45):28206-28213).

Immunogen

Native rabbit plasma CETP/LTP.

Application

Anti- LTP/CETP Antibody, clone 3-11D is an antibody against LTP/CETP for use in Western Blotting, ELISA, Inhibits Activity/Function.
ELISA Analysis: Representative lots were used as the capture antibody in combination with clone 14–8F as the detection antibody for the detection of human plasma CETP/LTP by sandwich ELISA (Tosheska, K., et al. (2011). Bosn. J. Basic. Med. Sci. 11(3):169–173; Saito, K., et al. J. Lipid Res. (1999). 40(11):2013-2021; Sasai, K., et al. (1998). Clin. Chem. 44(7):1466-1473).
ELISA Analysis: A representative lot was used as the capture antibody in combination with clone 14–8F as the detection antibody for the detection of mouse plasma CETP/LTP by sandwich ELISA (Wu, C.A., et al. (2002). Arterioscler .Thromb. Vasc. Biol. 22(8):1347-1353).
ELISA Analysis: A representative lot showed no significant pH-dependent CETP/LTP-capturing activity in the pH range from 5.0 to 7.5 in sandwich ELISA applications (Saito, K., et al. J. Lipid Res. (1999). 40(11):2013-2021).
Western Blotting Analysis: A representative lot detected only non-reduced, but not reduced, rabbit and human CETP/LTP by Western blotting (Ko, K.W., et al. (1994). J. Biol. Chem. 269(45):28206-28213).
Activity/Function Inhibition Analysis: A representative lot inhibited both rabbit and human CETP/LTP-mediated cholesteryl ester (CE) and triglyceride (TG) transfer from low density lipoprotein (LDL) to high density lipoprotein (HDL) (Ko, K.W., et al. (1994). J. Biol. Chem. 269(45):28206-28213).
Activity/Function Inhibition Analysis: A representative lot blocked the lipid association of both rabbit or human CETP/LTP with microemulsion (Ko, K.W., et al. (1994). J. Biol. Chem. 269(45):28206-28213).
Activity/Function Inhibition Analysis: A representative lot inhibited human CETP/LTP-mediated cholesteryl ester (CE) and triglyceride (TG) transfer from high density lipoprotein (HDL) to to apoB-containing lipoproteins in human plasma (Ko, K.W., et al. (1994). J. Biol. Chem. 269(45):28206-28213).
Research Category
Signaling
Research Sub Category
Lipid Metabolism & Weight Regulation

Quality

Evaluated by Western Blotting of purified human plasma CETP/LTP.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected purified human plasma CETP/LTP under non-reducing condition.

Target description

~70 kDa observed. Target band appears larger than the calculated molecular weight of 54.5 kDa due to glycosylation.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service