Skip to Content
Merck
All Photos(2)

Key Documents

MABN722

Sigma-Aldrich

Anti-WIF1 Antibody, clone 1G5

ascites fluid, clone 1G5, from mouse

Synonym(s):

WIF-1, WIF, Wnt inhibitory factor 1, WIF1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€404.00

€404.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€404.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€404.00


Estimated to ship on26 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1G5, monoclonal

species reactivity

mouse, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WIF1(11197)

General description

WIF1, also known as Wnt inhibitory factor 1 or WIF-1, is a secreted protein that binds to Wnt proteins and inhibits their activity. WNT proteins have been shown to act as an extracellular signaling molecules and may be involved in the control of embryonic development. WIf1 contains an N-terminal WIF domain and 5 epidermal growth factor (EGF)-like domains. It has been implied in recent sutyes that WIF1 expression is downregulated in many types of cancers.

Immunogen

Purified recombinant fragment of human WIF1 expressed in E. Coli.

Application

Anti-WIF1 Antibody, clone 1G5 is a highly specific mouse monoclonal antibody, that targets Wnt inhibitory factor 1 & has been tested in western blotting, IHC & Immunofluorescence.
Immunofluorescence Analysis: A 1:200-1,000 dilution from a representative lot detected WIF1 in HeLa cells.

Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected WIF1 in human ovarian tumor and lung cancer tissues.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in HeLa, NIH/3T3 and NTERA-2 cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected WIF1 in HeLa, NIH/3T3 and NTERA-2 cell lysates.

Target description

~43 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa, NIH/3T3 and NTERA-2 cell lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theresa A Reno et al.
BMC cancer, 16, 439-439 (2016-07-13)
The natural compound triptolide has been shown to decrease cell proliferation and induce apoptosis and cellular senescence. We previously demonstrated that triptolide decreases tumor formation and metastasis of human non-small cell lung cancer cells (NSCLC). Due to the toxicity of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service