Skip to Content
Merck
All Photos(2)

Key Documents

MABN704

Sigma-Aldrich

Anti-ASS1 Antibody, clone 2C10

clone 2C10, from mouse

Synonym(s):

Argininosuccinate synthase, Citrulline--aspartate ligase, ASS1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€404.00

€404.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€404.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€404.00


Estimated to ship on01 June 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2C10, monoclonal

species reactivity

mouse, human, rat, monkey

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ASS1(445)

General description

Argininosuccinate synthase (ASS1) or alternatively known by the older name of Citrulline-Aspartate ligase is encoded by the ASS1/ASS gene in humans and is the enzyme that is responsible for the third step in the urea cycle critical to the metabolism of nitrogen. ASS1 catalyzes the synthesis of argininosuccinate from citrulline and aspartate. Argininosuccinate is the immediate precursor of arginine and ASS1 was first identified as the rate limiting step in the liver for this reaction. ASS1 is now recognized to be expressed ubiquitously in mammalian tissues. ASS1 also performs the rate limiting step for NO production in the citrulline-NO cycle as well. Regulation of ASS1 expression can vary depending upon the tissue examined. In the liver, where arginine is hydrolyzed to form urea and ornithine, ASS1 is highly expressed, and various hormones such as glucocorticoids, glucagon, insulin and even high dietary levels of specific amino acids like glutamine regulate expression of ASS1 during development and adult life. In other tissues and in particular in nitric oxide (NO) producing cells, ASS1 is expressed at a low level but induced by proinflammatory cytokines. Interestingly ASS1 expression is also increased in Alzheimer’s disease in both neurons and glia, which also is correlated with iNOS expression and suggests a link between increased NO production and iNOS expression observed in Alzheimer brains. EMD-Millipore’s Anti-ASS1 monoclonal antibody has been tested in western blot on recombinant protein and cell lysates from A431, Raji, MOLT4, Jurkat, A549, NIH3T3, PC-12, and COS7 and in paraffin embedded immunohistochemistry on cervical cancer tissue sections. The antibody has also been tested in direct ELISA on recombinant antigen.

Immunogen

Purified recombinant fragment of human ASS1 expressed in E. Coli.

Application

Detect Argininosuccinate synthase using this mouse monoclonal antibody, Anti-ASS1 Antibody, clone 2C10 validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected ASS1 in human cervical cancer and colon cancer tissues.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Western Blotting in A431, Raji, MOLT4, Jurkat, A549, NIH/3T3, PC-12 and Cos7 cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected ASS1 in A431, Raji, MOLT4, Jurkat, A549, NIH/3T3, PC-12 and Cos7 cell lysates.

Target description

~47 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal in PBS with up to 0.1% sodium azide and 0.5% protein stabilizer.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431, Raji, MOLT4, Jurkat, A549, NIH/3T3, PC-12 and Cos7 cell lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah E Barnett et al.
Molecular cancer research : MCR, 21(5), 411-427 (2023-01-21)
The nuclear deubiquitylase BRCA1-associated protein 1 (BAP1) is frequently inactivated in malignant pleural mesothelioma (MPM) and germline BAP1 mutation predisposes to cancers including MPM. To explore the influence on cell physiology and drug sensitivity, we sequentially edited a predisposition mutation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service