Skip to Content
Merck
All Photos(2)

Key Documents

MABN69

Sigma-Aldrich

Anti-Caspr Antibody, clone K65/35

clone K65/35, from mouse

Synonym(s):

contactin associated protein 1, neurexin 4, neurexin-4, contactin-associated protein 1, Neurexin IV, Neurexin-4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€502.00

€502.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€502.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€502.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

K65/35, monoclonal

species reactivity

rat

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Contactin-associated protein 1(Caspr) is a single-pass type I membrane protein belonging to the neurexin family and is comprised of one F5/8 type C domain, one fibrinogen C-terminal domain, two EGF-like domains, and four laminin G-like domains. Caspr is well documented for its interaction with contactin, an interaction that is crucial to the transport of Caspr to the plasma membrane, the mylination of glial cells, and the generation of axoglial junctions. Studies also suggest that Caspr may play a regulatory role in the cell surface expression of contactin and its targeting to different axonal domains. Expression of Caspr is largely observed in the brain, mainly in CNS myelinated nerve fibers within paranodal axoglial junctions.

Immunogen

Recombinant protein corresponding to rat Caspr.

Application

Anti-Caspr Antibody, clone K65/35 is an antibody against Caspr for use in IH & WB.
Immunohistochemistry Analysis: 1:300 dilution from a previous lot detected Caspr in rat cerebellum and rat hippocampus tissues.
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Western Blot in rat brain membrane tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Caspr on 10 µg of rat brain membrane tissue lysate.

Target description

~ 220 kDa observed

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain membrane tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacob C A Edvinsson et al.
The journal of headache and pain, 23(1), 26-26 (2022-02-19)
5-Hydroxytryptamine (5-HT) receptors 1B, 1D and 1F have key roles in migraine pharmacotherapy. Selective agonists targeting these receptors, such as triptans and ditans, are effective in aborting acute migraine attacks and inhibit the in vivo release of calcitonin gene-related peptide
Anirban Paul et al.
Cell, 171(3), 522-539 (2017-09-26)
Understanding the organizational logic of neural circuits requires deciphering the biological basis of neuronal diversity and identity, but there is no consensus on how neuron types should be defined. We analyzed single-cell transcriptomes of a set of anatomically and physiologically characterized
Annika Balraj et al.
Developmental neurobiology, 82(4), 308-325 (2022-04-12)
Retinal ganglion cells generate a pattern of action potentials to communicate visual information from the retina to cortical areas. Myelin, an insulating sheath, wraps axonal segments to facilitate signal propagation and when deficient, can impair visual function. Optic nerve development
Aida Maddahi et al.
The journal of headache and pain, 23(1), 152-152 (2022-12-02)
Hypothalamus is a key region in migraine attacks. In addition, women are disproportionately affected by migraine. The calcitonin gene-related peptide (CGRP) system is an important key player in migraine pathophysiology. CGRP signaling could be a target of hormones that influence
Ana Palma et al.
Frontiers in cell and developmental biology, 10, 841548-841548 (2022-04-05)
Abnormalities in myelination are associated to behavioral and cognitive dysfunction in neurodevelopmental psychiatric disorders. Thus, therapies to promote or accelerate myelination could potentially ameliorate symptoms in autism. Clemastine, a histamine H1 antagonist with anticholinergic properties against muscarinic M1 receptor, is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service