Skip to Content
Merck
All Photos(1)

Key Documents

MABN683

Sigma-Aldrich

Anti-ETV5 Antibody, clone 3H3

ascites fluid, clone 3H3, from mouse

Synonym(s):

ETS translocation variant 5, Ets-related protein ERM, ETV5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€370.00

€370.00


Estimated to ship on08 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€370.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€370.00


Estimated to ship on08 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

3H3, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ETV5(2119)

General description

ETS family members play an important role in mammalian development and have been implicated in the regulation of gene expression during a variety of biologic processes including cell growth and differentiation. Defects or deficiencies, as well as overexpression of ETS family members can have major developmental effects as well as aid in tumor development. ETS translocation variant 5 protein (ETV5) also known as ETS-related protein ERM is a nuclear localized protein that binds to DNA sequences containing the consensus nucleotide core sequence of GGAA and is a member of the very large ETS transcription factor family. Early experiments in worms suggested that ETV5 might play a role in the development of the dopaminergic nervous system; however that has not proved to be the case in mammals. However, more recent research has indicated ETV5 playing an important role in the development of reproduction and in the development of ovarian cancer. ETV5 controls the expression of critical prostaglandins needed for ovarian development and ETV5 deficient mice show complex infertility phenotypes both in males and females. In males, the infertility phenotype exhibited by ETV5 deficiency is sexually dimorphic, and it involves both somatic cells and germ cells. In Etv5-/- female mice, the problem is more complicated by hormonal involvement and results in more a complex phenotype. EMD-Millipore’s Anti-ETV5 mouse monoclonal antibody has been tested in western blot against Jurkat, NIH3T3 and MCF-7 cell lysates.

Immunogen

Purified recombinant fragment of human ETV5 expressed in E. Coli.

Application

Anti-ETV5 Antibody, clone 3H3 is a highly specific mouse monoclonal antibody & that targets Ets & has been tested in western blotting.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in Jurkat, NIH/3T3 and MCF-7 cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected ETV5 in Jurkat, NIH/3T3 and MCF-7 cell lysates.

Optimal working dilutions must be determined by end user.

Target description

~58 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Jurkat, NIH/3T3 and MCF-7 cell lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Félicie Cottard et al.
Oncotarget, 8(42), 72008-72020 (2017-10-27)
Constitutively active androgen receptor (AR) variants have been involved in the expression of mesenchymal markers such as N-cadherin in prostate cancer (PCa). However, the underlying molecular mechanisms remain elusive. It remains unclear, whether N-cadherin gene (CDH2) is a direct transcriptional
Weiming Ouyang et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(8), 4078-4087 (2020-02-12)
Anthrax lethal toxin (LT) is a protease virulence factor produced by Bacillus anthracis that is required for its pathogenicity. LT treatment causes a rapid degradation of c-Jun protein that follows inactivation of the MEK1/2-Erk1/2 signaling pathway. Here we identify COP1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service