Skip to Content
Merck
All Photos(1)

Key Documents

MABN677

Sigma-Aldrich

Anti-ADNP1 Antibody, clone 5F5.1

clone 5F5.1, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€306.00

€306.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€306.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€306.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5F5.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ADNP(23394)

General description

ADNP1, also known as Activity-dependent neuroprotector homeobox protein, and encoded by the gene ADNP/ADNP1/KIAA0784, is a potential transcription factor that appears to mediate some of the neuroprotective peptide VIP associated effects involving normal growth and cancer proliferation. ADNP1 is widely expressed and localized to the nucleus. ADNP1 is strongly expressed in heart and skeletal muscle tissue and the brain. ADNP1 is also found expressed in colon and breast cancer tissues but not in normal tissues, thus ADNP1 can be used as a potential cancer marker in some tissues. ADNP1 is upregulated in normal proliferative tissues by VIP, and it may increase the viability of certain cell types through the modulation of p53 activity. Interestingly, new research shows that ADNP1 may be a potential biomarker for Alzheimer’s disease, where ADP1 expression was specifically down-regulated in AD patients.

Immunogen

Epitope: Near C-terminus
GST-tagged recombinant protein corresponding to human ADNP1, near the C-terminus.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-ADNP1 Antibody, clone 5F5.1 is validated for use in Western Blotting for the detection of ADNP1.

Quality

Evaluated by Western Blotting in SH-SY-5Y cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected ADNP1 in 200 µg of SH-SY-5Y cell lysate.

Target description

~145 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service