Skip to Content
Merck
All Photos(3)

Key Documents

MABN667

Sigma-Aldrich

Anti-TRPML3 Antibody, clone N268/18

clone N268/19, from mouse

Synonym(s):

Mucolipin-3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€421.00

€421.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€421.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€421.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N268/19, monoclonal

species reactivity

human, mouse, rat

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MCOLN3(55283)

General description

Mucolipin-3 (TRPML3) is a multi-pass membrane protein that functions as an ion channel. TRPML3 is possibly involved in locomotion and the differentiation of auditory receptor cells.

Specificity

This antibody recognizes the N-terminus of TRPML3.

Immunogen

Linear peptide corresponding to the N-terminus of human TRPML3.

Application

Detect Mucolipin-3 using this mouse monoclonal antibody, Anti-TRPML3 Antibody & clone N268/18 validated for use in western blotting.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected TRPML3 in 10 µg of mouse brain tissue lysate.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected TRPML3 in 10 µg of human brain tissue lysate.

Quality

Evaluated by Western Blotting in rat hippocampus tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected TRPML3 in 10 µg of rat hippocampus tissue lysate.

Target description

~ 64 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service