Skip to Content
Merck
All Photos(2)

Key Documents

MABN501

Sigma-Aldrich

Anti-SUR1 Antibody, clone N289/16

clone N289/16, from mouse

Synonym(s):

ATP-binding cassette sub-family C member 8, Sulfonylurea receptor 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€461.00

€461.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€461.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€461.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N289/16, monoclonal

species reactivity

human, mouse

species reactivity (predicted by homology)

rat (immunogen homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ABCC8(6833)

General description

Sulfonylurea receptor 1 (SUR1) is also called ATP-binding cassette sub-family C member 8, and is a member of the ABC transporter superfamily. SUR1 is a putative subunit of the beta-cell ATP-sensitive potassium channel (KATP) and functions as a regulator of ATP-sensitive K+ channels and insulin release.

Specificity

This antibody recognizes the C-terminus of SUR1. Representative lot deteted Sur1 in crude brain membrane preparations
in SUR1 wild-type (WT) mouse cell lysate, and demonstrated a loss of signal in SUR1 knockout (KO) mouse cell lysate (Prof. J. Trimmer, University of California, Davis.).

Immunogen

Recombinant protein corresponding to the C-terminus of rat SUR1.

Application

Anti-SUR1 Antibody, clone N289/16 is a highly specific mouse monoclonal antibody, that targets SUR & has been tested in western blotting & IHC.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected SUR1 in human thalamus tissue.

Quality

Evaluated by Western Blotting in mouse hypothalamus tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected SUR1 in 10 µg of mouse hypothalamus tissue lysate.

Target description

~ 180 kDa observed

Physical form

Format: Purified

Analysis Note

Control
Mouse hypothalamus tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ching-Han Lin et al.
The Journal of biological chemistry, 298(5), 101904-101904 (2022-04-11)
Pancreatic β-cells express ATP-sensitive potassium (KATP) channels, consisting of octamer complexes containing four sulfonylurea receptor 1 (SUR1) and four Kir6.2 subunits. Loss of KATP channel function causes persistent hyperinsulinemic hypoglycemia of infancy (PHHI), a rare but debilitating condition if not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service