Skip to Content
Merck
All Photos(1)

Key Documents

MABN385

Sigma-Aldrich

Anti-NHERF-2 Antibody, clone 32B6

clone 32B6, from mouse

Synonym(s):

Na(+)/H(+) exchange regulatory cofactor NHE-RF2, NHERF-2, NHE3 kinase A regulatory protein E3KARP, SRY-interacting protein 1, SIP-1, Sodium-hydrogen exchanger regulatory factor 2, Solute carrier family 9 isoform A3 regulatory factor 2, Tyrosine kinase ac

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€316.00

€316.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€316.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€316.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

32B6, monoclonal

species reactivity

rat, human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLC9A3R2(9351)

General description

Na(+)/H(+) exchange regulatory cofactor NHE-RF2 (NHERF-2) is also known as NHE3 kinase A regulatory protein E3KARP, SRY-interacting protein 1 (SIP-1), Sodium-hydrogen exchanger regulatory factor 2, Solute carrier family 9 isoform A3 regulatory factor 2 (SLC9A3R2), and Tyrosine kinase activator protein 1 (TKA-1). NHERF-2 is a Scaffold protein that links ezrin/moesin/radixin family members to the actin cytoskeleton and regulates their surface expression. NHERF-2 is critical for the cAMP-mediated phosphorylation and inhibition of SLC9A3. NHERF-2 is also thought to act as scaffold protein in the nucleus.

Immunogen

recombinant protein corresponding to human NHERF-2.

Application

Anti-NHERF-2 Antibody, clone 32B6 is a highly specific mouse monoclonal antibody, that targets NHERF2 & has been tested in western blotting, ICC & IP.
Immunocytochemistry Analysis: A representative lot was used by an independent laboratory in A431 and NHERF-2 transfected HT1080 cells. (Theisen, C. S., et al. (2007). Mole. Biol. Cell. 18:1220-1232).
Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in Batt cell lysate. (Theisen, C. S., et al. (2007). Mole. Biol. Cell. 18:1220-1232).
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in HaCaT keratinocytes cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected NHERF-2 in 10 µg of HaCaT keratinocytes cell lysate.

Target description

~ 37 kDa observed. An uncharacterized band may appear at ~41 kDa in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HaCaT keratinocytes cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service