Skip to Content
Merck
All Photos(2)

Key Documents

MABN296

Sigma-Aldrich

Anti-RLBP1 Antibody, clone 3C1.1

clone 3C1.1, from mouse

Synonym(s):

Anti-Retinaldehyde-binding protein 1, Cellular retinaldehyde-binding protein, RLBP1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€408.00

€408.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€408.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€408.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C1.1, monoclonal

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RLBP1(6017)

General description

Retinaldehyde-binding protein 1 (RLBP1), also known as Cellular retinaldehyde-binding protein, and encoded by the gene RLBP1/CRALBP, is a soluble retinoid carrier protein essential for the proper function of both rod and cone photoreceptors. RLBP1 participates in the regeneration of active 11-cis-retinol and 11-cis-retinaldehyde from the inactive 11-trans products of the rhodopsin photocycle and in the de novo synthesis of these retinoids from 11-trans metabolic precursors. The cycling of retinoids between photoreceptor and adjacent pigment epithelium cells is known as the “visual cycle”. RLBP1 is localized to the cytoplasm. RLBP1 is expressed in retina and pineal gland. It not present in photoreceptor cells but is expressed abundantly in the adjacent retinal pigment epithelium (RPE) and in the Mueller glial cells of the retina. Defects in RLBP1 may be associated with Retinitis pigmentosa, Bothnia retinal dystrophy (BRD), Rod-cone dystrophy Newfoundland, and Retinitis punctata albescens genetic diseases.

Immunogen

GST-tagged recombinant protein corresponding to human RLBP1.

Application

Anti-RLBP1 Antibody, clone 3C1.1 is a highly specific mouse monoclonal antibody, that targets Retinaldehyde-binding protein 1 & has been tested in western blotting & IHC.
Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected RLBP1 in human retina and rat retina tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in rat eye tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected RLBP1 in 10 µg of rat eye tissue lysate.

Target description

~37 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service