MABN2621
Anti-UNC119 homolog A/mRG4 Antibody, clone A2
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
A2, monoclonal
mol wt
calculated mol wt 27.01 kDa
observed mol wt ~28 kDa
purified by
using protein G
species reactivity
mouse
packaging
antibody small pack of 100 μg
technique(s)
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
isotype
IgG1κ
epitope sequence
Unknown
Protein ID accession no.
UniProt accession no.
shipped in
2-8°C
target post-translational modification
unmodified
Gene Information
mouse ... Unc119(22248)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in Mouse retina tissue lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected UNC119 homolog A (mRG4) in Mouse retina tissue lysate.
Tested Applications
Immunofluorescence Analysis: A representative lot detected UNC119 homolog A/mRG4 in Immunofluorescence applications (Haeseleer, F., et al. (2008). Invest Ophthalmol Vis Sci. 49(6):2366-75).
Western Blotting Analysis: A representative lot detected UNC119 homolog A/mRG4 in Western Blotting applications (Haeseleer, F., et al. (2008). Invest Ophthalmol Vis Sci. 49(6):2366-75).
Immunohistochemistry Applications: A representative lot detected UNC119 homolog A/mRG4 in Immunohistochemistry applications (Haeseleer, F., et al. (2008). Invest Ophthalmol Vis Sci.;49(6):2366-75).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service