Skip to Content
Merck
All Photos(1)

Key Documents

MABF714

Sigma-Aldrich

Anti-CD54 (ICAM-1) Antibody (human), PE, clone 15.2

clone 15.2, from mouse

Synonym(s):

Intercellular adhesion molecule 1, ICAM-1, Major group rhinovirus receptor, CD antigen CD54

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€260.00

€260.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 TESTS
€260.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€260.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified antibody

antibody product type

primary antibodies

clone

15.2, monoclonal

species reactivity

human

packaging

pkg of 5 μL (0.5 μg/test)

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ICAM1(3383)

General description

The 15.2 antibody reacts with human CD54, also known as ICAM-1 (Intercellular Adhesion Molecule 1), a 90-110 kDa cell surface glycoprotein that is inducibly expressed on both immune and endothelial cells. As its name implies, ICAM-1 participates in cell-cell adhesion between leukocytes and endothelial cells, facilitating leukocyte recruitment and transmigration at sites of inflammation. The ligands for ICAM-1 are also expressed on leukocyte and endothelial cells, and include Mac-1, fibrinogen, and a member of the integrin protein family, LFA-1 (CD11a).The 15.2 antibody may be used for analysis of ICAM-1 expression in human cells and tissues, and is reported to be cross-reactive with porcine ICAM-1.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
This mouse monoclonal antibody Anti-CD54 (ICAM-1) (human), PE, clone 15.2 validated for use in western blotting, IHC & Flow Cytometry for the detection of CD54 (ICAM-1).

Quality

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.5 µg of this antibody detected CD54 (ICAM-1) in one million human peripheral blood monocytes.

Physical form

Purified Mouse Monoclonal Antibody in buffer containing 10 mM NaH2PO4, 150 mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Size Exclusion Chromatography

Storage and Stability

It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service