Skip to Content
Merck
All Photos(1)

Key Documents

MABF508

Sigma-Aldrich

Anti-CD115 (c-fms) Antibody (mouse), PE, clone AFS98

clone AFS98, 0.2 mg/mL, from rat

Synonym(s):

Macrophage colony-stimulating factor 1 receptor, CSF-1 receptor, CSF-1-R, CSF-1R, M-CSF-R, Proto-oncogene c-Fms, CD115, CSF1-R

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€152.00

€152.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€152.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€152.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rat

Quality Level

conjugate

PE

antibody form

purified antibody

antibody product type

primary antibodies

clone

AFS98, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

concentration

0.2 mg/mL

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Csf1R(12978)

Related Categories

General description

The AFS98 antibody is specific for mouse CD115, also known as Colony-Stimulating Factor-1 Receptor (CSF-1R), a 145 kDa receptor from the PDGF receptor family. Receptor activation by the ligands IL-34 or CSF-1 (M-CSF) occurs via homodimerization of CD115 and subsequent tyrosine phosphorylation and ubiquitination of intracellular domains. CD115 signaling promotes differentiation of myeloid precursors, as well as the continued regulation of proliferation, survival and function of mononuclear phagocytes, dendritic cells and osteoclasts. While IL-34 and CSF-1 may induce similar cellular responses, they are differentially expressed and as such exert complimentary actions via CD115.The AFS98 antibody may be used for identification of myeloid lineage cells by flow cytometry, and is commonly used for in vivo or in vitro neutralization of CSF-1 Receptor.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
This rat monoclonal antibody Anti-CD115 (c-fms) (mouse), PE, clone AFS98 validated for use in western blotting & Flow Cytometry for the detection of CD115 (c-fms).

Quality

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.125 µg of this antibody detected CD115 (c-fms) in one million C57Bl/6 peripheral blood cells.

Physical form

Purified Rat Monoclonal Antibody in buffer containing 10 mM NaH2PO4, 150 mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Size Exclusion Chromatography

Storage and Stability

It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service