Skip to Content
Merck
All Photos(1)

Key Documents

MABF41

Sigma-Aldrich

Anti-Interleukin-6 (IL-6) Antibody, clone 1G3

clone AB42_51.38C8.1G3, from mouse

Synonym(s):

interleukin 6 (interferon, beta 2), IFN-beta-2, B-cell stimulatory factor 2, interleukin-6, Interferon beta-2, B cell stimulatory factor-2, Hybridoma growth factor, B-cell differentiation factor, CTL differentiation factor, interleukin BSF-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€324.00

€324.00


Estimated to ship on24 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1982

Request a Bulk Order

Select a Size

Change View
100 μG
€324.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€324.00


Estimated to ship on24 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1982

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

AB42_51.38C8.1G3, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Interleukin-6 (IL-6) is a pro-inflammatory cytokine that seems to be involved in the late stages of CTL generation. Other biological functions also include amplification and the inflammatory response. Combination therapies including IL-6 have been shown to have positive effects on specific malignant tumor types. The accumulation of IL-6 in the CNS is usually detrimental and can contribute to pathology of CNS diseases. IL-6 signaling is negatively regulated by TGF-beta, which down regulates STAT1, STAT3 and ICAM-1 expression. IL-6 expression has been linked to inflammatory bowel disease (IBD) and is also the primary clinical target for IBD. Clinical use of an antibody for the IL-6 receptor (IL-6R) has been promising in treating rheumatoid arthritis, systemic juvenile idiopathic arthritis and Castleman’s disease.

Immunogen

Human IL-6 fused to the soluble domain of the IL-6R.

Application

Anti-Interleukin-6 (IL-6) Antibody, clone 1G3 is an antibody against Interleukin-6 (IL-6) for use in WB.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors

Quality

Evaluated by Western Blot with human IL-6 recombinant protein.

Western Blot Analysis: 0.5 g/mL of this antibody detected 10 g of human IL-6 recombinant protein.

Target description

~21 kDa observed

Linkage

Replaces: 04-1095

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human IL-6 recombinant protein

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoli Zhou et al.
Journal of the American Society of Nephrology : JASN, 27(6), 1765-1776 (2015-10-17)
Deficiency of the antiaging gene Klotho (KL) induces renal damage and hypertension through unknown mechanisms. In this study, we assessed whether KL regulates expression of CYP11B2, a key rate-limiting enzyme in aldosterone synthesis, in adrenal glands. We found that haplodeficiency
Yan Wang et al.
Experimental and therapeutic medicine, 15(3), 2830-2836 (2018-02-20)
The natural iso-flavonoid, biochanin A, is categorized as a phytoestrogen and has been demonstrated to exhibit various pharmacological properties. However, no effects of biochanin A on lung cancer cell lines have been reported. Therefore, the present study aimed to demonstrate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service