Skip to Content
Merck
All Photos(2)

Key Documents

MABF3181

Sigma-Aldrich

Anti-IL-4 Antibody, clone 2D7.14.8

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€146.00
100 μG
€370.00

€146.00


Estimated to ship on24 May 2025



Select a Size

Change View
25 μG
€146.00
100 μG
€370.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€146.00


Estimated to ship on24 May 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2D7.14.8, monoclonal

mol wt

calculated mol wt 17.49 kDa
observed mol wt ~15 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... IL4(3565)

General description

Interleukin-4 (UniProt: P05112; also known as IL-4, B-cell stimulatory factor 1, BSF-1, Binetrakin, Lymphocyte stimulatory factor 1, Pitrakinra) is encoded by the IL4 gene (Gene ID: 3565) in human. IL-4 is a cytokine that is secreted primarily by mast cells, Th2 cells, eosinophils, and basophils. IL-4 is synthesized with a signal peptide (1-24), which is subsequently cleaved off to produce the mature form. It participates in the activation of B cells and other cell types. It also serves as a co-stimulator of DNA-synthesis. It induces the expression of class II MHC molecules on resting B-cells. It enhances both secretion and cell surface expression of IgE and IgG1. It also reported to regulate the expression of the low affinity Fc receptor for IgE (CD23) on both lymphocytes and monocytes. IL-4 inhibits macrophage production of proinflammatory cytokines, including TNF-a, IL-1, and IL-6 via transcriptional repression. It also enhances production of molecules that counteract these cytokines, such as IL-1R antagonist and the decoy receptor IL-1RII. The effects of IL-4 signaling are mediated through binding to the IL-4 receptor a chain (IL-4R ) that dimerizes with g-chain upon ligand binding to produce type I signaling complex located mainly of hematopoietic cells. It can also dimerize with IL-13a receptor a (IL-13Ra1) to produce type-2 complex that is expressed also on non-hematopoietic cells. IL-4 is also shown to stimulate autophagy in dendritic cells by interfering with mTORC1 signaling through the induction of RUN and FYVE domain containing 4 (RUFY4). (Ref.: Gadani, SP., et al. (2012). J. Immunol. 189(9); 4213-4219; Otsuka, TY., et al. (1986). Proc. Natl. Acad. Sci. USA. 83(16); 5894-5898).

Specificity

Clone 2D7.14.8 is a mouse monoclonal antibody that detects human Interleukin-4.

Immunogen

Full-length recombinant human Interleukin-4 (without the signal peptide).

Application

Quality Conrol Testing

Evaluated by Western Blotting with recombinant human IL-4

Western Blotting Analysis (WB): A 1:10,000 dilution of this antibody detected recombinant human IL-4.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected IL-4 in human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-IL-4, clone 2D7.14.8, Cat. No. MABF3181, is a mouse monoclonal antibody that detects human Inerleukin-4 and is is tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG2a in PBS with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service