Skip to Content
Merck
All Photos(1)

Documents

MABF3141

Sigma-Aldrich

Anti-ANTXR2/CMG-2 Antibody, clone 1H8

Synonym(s):

Anthrax toxin receptor 2, CMG-2, Capillary morphogenesis gene 2vprotein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1H8, monoclonal

species reactivity

human, mouse

packaging

antibody small pack of 100

technique(s)

western blot: suitable

isotype

IgG2cκ

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

Gene Information

human ... ANTXR2(118429)

Specificity

Clone 1H8 is a mouse monoclonal antibody that detects Anthrax toxin receptor 2 (ANTXR2). It targets an epitope within the N-terminal half.

Immunogen

A recombinant fragment of 178 amino acids from the N-terminal half of human ANTXR2 with an N-terminal extension of GSHMLEDPRG.

Application

Quality Control Testing

Evaluated by Western Blotting in Mouse lung tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ANTXR2/CMG-2 in Mouse lung tissue lysate.

Tested Applications

Western Blotting Analysis: A representative lot detected ANTXR2/CMG-2 in Western Blotting applications (Chaudhary, A., et al. (2012). Cancer Cell. 21(2):212-26; Szot, C., et al. (2018). J Clin Invest. 128(7):2927-2943).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Anthrax toxin receptor 2 (UniProt: P58335; also known as Capillary morphogenesis gene 2 protein, CMG-2) is encoded by the ANTRX2 (also known as CMG2) gene (Gene ID: 118429) in human. ANTRX2/CMG-2 is widely expressed, single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-33), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 34-318), a transmembrane domain (aa 319-341), and a cytoplasmic domain (aa 342-489). It shares about 58% amino acid identity with ANTXR1. Four isoforms of ANTRX2/CMG-2 have been described that are produced by alternative splicing. It is reported to be essential for cellular interactions with laminin and the extracellular matrix. It is an important regulator of collagen VI homeostasis. It mediates the transport of collagen to lysosomes for degradation. Its loss promotes accumulation of Collagen VI leading to nodule formation. Antrx2 -/- mice are shown to display massive accumulation of collagen VI in their uteri leading to fibrosis and sterility. It binds to collagen VI via its vWA domain (aa 44-213) in a MIDAS (metal ion-dependent binding domain) manner. (Ref.: Burgi, J., et al. (2017). Nat. Commun. 8; Article 15861; Chaudhary, A., et al. (2012). Cancer Cell. 21(2): 212-226).

Physical form

Purified mouse monoclonal antibody IgG2c in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service