Skip to Content
Merck
All Photos(1)

Key Documents

MABF272

Sigma-Aldrich

Anti-TREM-1, clone 2(1a2) Antibody

clone 2(1a2), from mouse, Atto 550 conjugate

Synonym(s):

Triggering receptor expressed on myeloid cells 1, TREM-1, Triggering receptor expressed on monocytes 1, CD354

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€315.00

€315.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€315.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€315.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

Atto 550 conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2(1a2), monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TREM1(54210)

General description

Triggering receptor expressed on myeloid cells 1 (TREM-1) is a membrane bound protein and secreted protein that is expressed by most cells in many tissues including liver, lung, spleen, lymph node, heart, and even spinal cord in response to bacterial and fungal infections. TREM-1 stimulates neutrophil and monocyte mediated inflammatory responses. TREM-1 also triggers pro-inflammatory chemokines and cytokines as well as increases surface expression of cell activation markers like CD11, thus TREM-1 acts as an amplifier of inflammatory responses in response to these pathogens. TREM-1 similarly is a potent LPS response factor and its expression is upregulated by LPS released by bacterial and fungal infections, and thus TREM-1 is a critical mediator of septic shock caused by such infections.

Immunogen

Recombinant protein corresponding to human TREM-1.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-TREM-1, clone 2(1a2) Antibody is validated for use in Flow Cytometry for the detection of TREM-1.

Quality

Evaluated by Flow Cytometry in human peripheral blood mononuclear.

Flow Cytometry Analysis: 0.25 µg of this antibody detected TREM-1 in human peripheral blood mononuclear cells.

Target description

26 kDa calculated

Physical form

Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service