Skip to Content
Merck
All Photos(1)

Key Documents

MABF233

Sigma-Aldrich

Anti-HLA-C Antibody, clone DT9

clone DT9, from mouse

Synonym(s):

HLA class I histocompatibility antigen,Cw-1 alpha chain, MHC class I antigen Cw*1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€418.00

€418.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€418.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€418.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

DT9, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2bκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HLA-C(3107)

Related Categories

General description

HLA-C, also known as HLA class I histocompatibility antigen, Cw-1 alpha chain, also known as MHC class I antigen Cw*1, and encoded by the gene HLA-C/HLAC. It contains an alpha and beta (beta-2-microglobulin) chain and interacts with the human herpesvirus 8 MIR1 protein.

The previously assigned protein identifier P30499 has been merged into P10321. Full details can be found on the UniProt database.

Immunogen

Purified tamarin MHC class I molecules corresponding to human HLA-C.

Application

Anti-HLA-C Antibody, clone DT9 is an antibody against HLA-C for use in flow cytometry & IP.
Flow Cytometry Analysis: A representative lot from an independent laboratory detected HLA-C in peripheral blood CD3+ cells from 200 healthy human donors (Apps, R., et al. (2013). Science. 340(6128):84-91.).

Flow Cytometry Analysis: A representative lot from an independent laboratory detected HLA-C in human PBMCs (Corrah, T. W., et al. (2011). J Virol. 85(7):3367-3374.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated HLA-C from various human cell lines (Braud, V. M., et al. (1998). Curr Biol. 8(1):1-10.).
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms

Quality

Evaluated by Flow Cytometry in human PBMCs.

Flow Cytometry Analysis: 1 µg of this antibody detected HLA-C in 1X10E6 human PBMCs.

Target description

41 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Etikala Apoorva et al.
Helicobacter, 29(2), e13069-e13069 (2024-03-22)
Helicobacter pylori (H. pylori) seems to play causative roles in gastric cancers. H. pylori has also been detected in established gastric cancers. How the presence of H. pylori modulates immune response to the cancer is unclear. The cytotoxicity of natural
Bizhan Romani et al.
Scientific reports, 7(1), 87-87 (2017-03-10)
Functional studies of HIV-1 proteins are normally conducted using lab adapted strains of HIV-1. The extent of those functions in clinical strains is sometimes unknown. In this study, we amplified and sequenced HIV-1 Vpu from 10 Iranian patients infected with
Bizhan Romani et al.
Scientific reports, 7, 44894-44894 (2017-03-21)
Functional studies of HIV-1 proteins are normally conducted using lab adapted strains of HIV-1. The extent of those functions in clinical strains is sometimes unknown. In this study, we amplified and sequenced HIV-1 Vpu from 10 Iranian patients infected with
Kristina Hopfensperger et al.
mBio, 11(4) (2020-07-16)
HLA-C-mediated antigen presentation induces the killing of human immunodeficiency virus (HIV)-infected CD4+ T cells by cytotoxic T lymphocytes (CTLs). To evade killing, many HIV-1 group M strains decrease HLA-C surface levels using their accessory protein Vpu. However, some HIV-1 group
Eri Yamada et al.
Cell host & microbe, 23(1), 110-120 (2018-01-13)
The HIV-1-encoded accessory protein Vpu exerts several immunomodulatory functions, including counteraction of the host restriction factor tetherin, downmodulation of CD4, and inhibition of NF-κB activity to facilitate HIV-1 infection. However, the relative contribution of individual Vpu functions to HIV-1 infection

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service