Skip to Content
Merck
All Photos(1)

Key Documents

MABF2225

Sigma-Aldrich

Anti-HSV-2 VP16 Antibody, clone LP1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€146.00
100 μL
€370.00

€146.00


Estimated to ship on21 May 2025



Select a Size

Change View
25 μL
€146.00
100 μL
€370.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€146.00


Estimated to ship on21 May 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

LP1, monoclonal

mol wt

calculated mol wt 54.6 kDa
observed mol wt ~37 kDa

purified by

using protein G

species reactivity

herpes simplex virus 1

packaging

antibody small pack of 100 μL

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

epitope sequence

Unknown

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

General description

Alpha trans-inducing protein (UniProt: Q69113; also known as Alpha-TIF) is encoded by the VP16 (also known as UL48) gene (Gene ID: 1487335) in human Herpes Simplex virus 2. Herpes simplex virus (HSV) is a large enveloped DNA virus that replicates in the nuclei of mammalian cells. Its virion contains two regulatory proteins that are reported to modulate gene expression in infected cells: the transcriptional activator VP16 and the virion host shutoff protein vhs. VP16, an abundant structural protein, is shown to be essential for virion assembly and it stimulates transcription of the HSV immediate-early (IE) genes. Its transcriptional activation domain is localized in the C-terminal region. It is targeted to the TAATGARATTC consensus sequence found in IE promoters through interactions with the host factors Oct-1 and HCF. It is delivered by the infecting virion and acts during the earliest stages of infection to stimulate transcription of the viral IE genes, thereby facilitating the onset of the lytic program of viral gene expression. It is also reported to suppress vhs host protein synthesis and induce accelerated turnover of cellular and viral mRNAs. Clone LP1 is directed against VP16 in both HSV-1 and HSV-2. (Ref.: Smibert, CA., et al. (1994). J. Virol. 68(4); 2339-2346; McLean, C., et al. (1982). J. Gen. Virol. 63(2); 297-305).

Specificity

Clone LP1 is a mouse monoclonal antibody that detects VP16 protein of Herpes simplex virus 2 (HSV-2) and displays cross reactivity with HSV-1.

Immunogen

Herpes simplex virus -2; strain 25766.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HSV-1 infected Vero cells.

Western Blotting Analysis (WB): A 1:500 dilution of this antibody detected VP16 protein in lysate from HSV-1 infected Vero cells.

Tested Applications

Immunoprecipitation Analysis): A representative lot immunoprecipitated HSV-2 VP16 in Immunoprecipitation applications (McLean, C., et al. (1982). J Gen Virol.; 63(2):297-305).

Western Blotting Analysis: A representative lot detected HSV-2 VP16 in Western Blotting applications (Crameri, M., et al. (2018). Nat Commun.; 9(1):1980; McLean, C., et al. (1982). J Gen Virol.; 63(2):297-305).

Immunofluorescence Analysis: A representative lot detected HSV-2 VP16 in Immunofluorescence applications (McLean, C., et al. (1982). J Gen Virol.; 63(2):297-305).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-HSV-2 VP16, clone LP1, Cat. No. MABF2225, is a mouse monoclonal antibody that detects Alpha trans-inducing protein (VP16) and is tested for use in Immunofluorescence, Immunoprecipitation, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service