Skip to Content
Merck
All Photos(2)

Key Documents

MABF215

Sigma-Aldrich

Anti-NOD1/CARD4 Antibody, clone 021

clone 021, 1 mg/mL, from mouse

Synonym(s):

Nucleotide-binding oligomerization domain-containing protein 1, Caspase recruitment domain-containing protein 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€343.00

€343.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€343.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€343.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

021, monoclonal

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NOD1(10392)

Related Categories

General description

NOD1/CARD4, also known as Nucleotide-binding oligomerization domain-containing protein 1, or Caspase recruitment domain-containing protein 4, and encoded by the gene NOD1/CARD4, is an important apoptosis mediating protein that enhances caspase-9-mediated apoptosis. NOD1/CARD4 also mediates cellular responsiveness to intracellular bacterial LPS and forms an intracellular sensing system along with another protein called ARHGEF2 for the detection of microbial effectors during cell invasion by pathogens. NOD1/CARD4 is a homodimer that is found in the cytoplasm but also associated with the cell membrane in particular cells such as polarized epithelial cells. NOD1/CARD4 is expressed in many tissues including heart, skeletal muscle, and pancreas. As part of the innate immune system NOD1/CARD4 is part of a family of proteins that are responsible for the initial response of an organism to potentially harmful stressors, pathogens or tissue injury, and accordingly plays an essential role in the pathogenesis of many inflammatory processes, including some cardiovascular diseases.

Immunogen

Epitope: CARD domain
Recombinant protein corresponding to the CARD domain of human NOD1/CARD4.

Application

Anti-NOD1/CARD4 Antibody, clone 021 is an antibody against NOD1/CARD4 for use in western blotting & flow cytometry.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Target description

~125 kDa observed. The calculated molecular weight of this protein is 108 kDa, but has been observed at ~125 kDa in some cell lysates. An IgG heavy chain may be observed at 55 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service