Skip to Content
Merck
All Photos(2)

Key Documents

MABF213

Sigma-Aldrich

Anti- STING, clone 41 Antibody

clone 41, 0.5 mg/mL, from rat

Synonym(s):

Stimulator of interferon genes protein, mSTING, Endoplasmic reticulum interferon stimulator, ERIS, Mediator of IRF3 activation, MMITA, Transmembrane protein 173, STING

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€381.00

€381.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€381.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€381.00


Check Cart for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

41, monoclonal

species reactivity

mouse

concentration

0.5 mg/mL

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Sting1(72512)

Related Categories

General description

STING, also known as mSTING, or Endoplasmic reticulum interferon stimulator, ERIS, Mediator of IRF3 activation, MMITA or Transmembrane protein 173, and encoded by the gene Tmem 173/Eris/Mita/Mpys/Sting, is a facilitator of innate immune signaling that acts as a sensor of cytosolic DNA from bacteria and viruses and promotes the production of type I interferons. STING acts by recognizing and binding cyclic di-GMP (c-di-GMP), a second messenger produced by bacteria, and cyclic GMP-AMP (cGAMP), a messenger produced in response to DNA virus in the cytosol. Upon binding STING is able to activate both NF-kappa-B and IRF3 transcription pathways to induce expression of type I interferon and exert a potent anti-viral state. STING also mediates cell death signaling via the activation of the ERK pathway. STING is found in multiple intracellular locations including the endoplasmic reticulum, plasma cell membrane, mitochondrion outer membrane, and perinuclear regions. STING is expressed in the B-cell lineage both immature and mature B cells but not plasma cells. Mutations in STING cause defects in innate immunity and compromised immune health.

Immunogen

Recombinant protein corresponding to the cytoplasmic domain of mouse STING.

Application

Flow Cytometry Analysis: 2 µg of this antibody detected STING in mouse splenocyte cells.
This Anti- STING, clone 41 Antibody is validated for use in Western Blotting and Flow Cytometry for the detection of STING.

Quality

Evaluated by Western Blotting in RAW264.7 cell lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected STING in 10 µg of RAW264.7 cell lysate.

Target description

~42 kDa observed. Uniprot describes 2 isoforms produced by alternative splicing at ~38 kDa and ~48 kDa

Physical form

Format: Purified

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manish Chauhan et al.
Cell death & disease, 15(3), 195-195 (2024-03-09)
STING (STimulator of Interferon Genes) is a cytosolic sensor for cyclic dinucleotides (CDNs) and initiates an innate immune response upon binding to CDNs. Coxiella burnetii is a Gram-negative obligate intracellular bacterium and the causative agent of the zoonotic disease Q
Kevin MingJie Gao et al.
Cell reports, 43(4), 114114-114114 (2024-04-16)
Patients afflicted with Stimulator of interferon gene (STING) gain-of-function mutations frequently present with debilitating interstitial lung disease (ILD) that is recapitulated in mice expressing the STINGV154M mutation (VM). Prior radiation chimera studies revealed an unexpected and critical role for non-hematopoietic
Ezra B Mead et al.
Insects, 15(6) (2024-06-26)
The arbovirus West Nile virus (WNV) is a danger to global health. Spread primarily by mosquitoes, WNV causes about 2000 cases per year in the United States. The natural mosquito immune response controls viral replication so that the host survives
Assaf Marcus et al.
Immunity, 49(4), 754-763 (2018-10-18)
Detection of cytosolic DNA by the enzyme cGAS triggers the production of cGAMP, a second messenger that binds and activates the adaptor protein STING, which leads to interferon (IFN) production. Here, we found that in vivo natural killer (NK) cell killing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service