Skip to Content
Merck
All Photos(2)

Documents

MABF2051

Sigma-Aldrich

Anti-Chikungunya virus Antibody, clone 6A11

clone 6A11, from mouse

Synonym(s):

CHIKV, E1 Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6A11, monoclonal

species reactivity

virus

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

target post-translational modification

unmodified

General description

Chikungunya virus (CHIKV) that causes distinctive polyarthritis or polyarthralgia with clinical features such as fever, maculopapular rash, and myalgia is transmitted by Aedes spp of mosquitoes. Immune compromised individuals may face serious complications, including encephalitis and mortality. CHIKV genome contains a single-stranded positive-sense RNA that encodes four non-structural proteins known as nsP1, nsP2, nsP3, and nsP4 and also five structural proteins that include a capsid protein, three envelope glycoproteins known as (E1, E2, and E3, and a small molecule known as 6K. The mature alphavirus particles express E1 and E2 heterodimers that form 80 trimeric spikes on the surface of the virion. The ectodomain E1 protein consists of three domains known as D1, DII, and DIII. The DIII domain is an immunoglobulin-like domain connected to D1 and DII by a flexible linker. Both E1 and E2 proteins are responsible for virus entry into host cells. The E2 glycoprotein interacts with a cellular receptor, resulting in the virus internalization and the E1 glycoprotein mediates virus fusion to host cell under low pH conditions. Following the fusion of the viral envelope with the endosomal membrane, the viral genomic RNA is released into the cytoplasm and starts replicating. (Ref.: Marsinoul, P., et al. (2014). Virology 464-465; 111-117; Lam, S., et al. (2015). mAbs 7(6), 1178-1194).

Specificity

Clone 6A11 specifically detects Chikungunya virus in infected cells.

Immunogen

CH122508 strain of Chikungunya virus.

Application

Anti-Chikungunya virus, clone 6A11, Cat. No. MABF2051 is a mouse monoclonal antibody that detects E1 protein of Chikungunya virus (CHIKV) and has been tested for use in Immunocytochemistry and Western Blotting.
Western Blotting Analysis: A 1:100 dilution from a representative lot detected Chikungunya virus in BHK cells infected with CHIKV (Courtesy of an outside independent laboratory).

Immunocytochemistry Analysis: A representative lot detected Chikungunya virus in BHK cells infected with CHIKV (Courtesy of an outside independent laboratory).

Quality

Isotype testing: Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG2b .

Target description

~9.17 kDa calculated for E1 protein. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L G Webb et al.
PLoS pathogens, 16(10), e1008999-e1008999 (2020-10-16)
Chikungunya virus (CHIKV) is a mosquito-borne alphavirus known to cause epidemics resulting in predominantly symptomatic infections, which in rare cases cause long term debilitating arthritis and arthralgia. Significant progress has been made in understanding the roles of canonical RNA sensing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service