Skip to Content
Merck
All Photos(1)

Key Documents

MABF2045

Sigma-Aldrich

Anti-Siglec-H Antibody, clone 551

clone 551, from rat

Synonym(s):

Sialic acid binding Ig-like lectin H, Siglech

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€154.00
100 μG
€418.00

€154.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€154.00
100 μG
€418.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€154.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

551, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Siglec-H (UniProt: Q3Y597; also known as Sialic acid binding Ig-like lectin H) is encoded by the Siglech gene (Gene ID: 233274) in murine species. Siglecs are sialic acid-binding immunoglobulin-like lectins that are expressed mostly in the immune system and may be involved in both adhesive and signaling functions. Siglec-H is a type I, CD33-related siglec-like transmembrane glycoprotein that is synthesized with a signal peptide (aa 1-18), which is subsequently cleaved off in the mature form. The mature form has to ig-like domains (aa 32-141 and 151-234) in its extracellular region. Unlike other CD33-related siglecs, it lacks tyrosine-based signaling motifs in its cytoplasmic tail. Siglec-H is shown to be expressed on plasmaocytoid dendritic cells (pDCs) and can serve as a reliable marker for these cells. Siglec-H is also reported to serve as an endocytic receptor expressed on murine pDCs. Action of Siglec-H depends on a 12 kDa DNAX-activating protein (DAP). (Ref.: Zhang, J., et al. (2006). Blood 107(9); 3600-3608; Yun, TJ., et al., (2016), Cell Metabolism 23(5); 852-866).

Specificity

Clone 551 is a rat monoclonal antibody that detects murine Siglec-H. It targets an epitope with in the extracellular domain.

Immunogen

Bone marrow-derived Type 1 interferon-producing cells.
Epitope: extracellular domain

Application

Anti-Siglec-H, clone 551, Cat. No. MABF2045, is a rat monoclonal antibody that detects Sialic acid binding Ig-like lectin H and has been tested for use in Flow Cytometry and Immunohistochemistry.
Immunohistochemistry Analysis: A representative lot detected Siglec-H in Immunohistochemistry applications (Yun, T.J., et. al. (2016). Cell Metab. 23(5):852-66).

Flow Cytometry Analysis: A representative lot detected Siglec-H in Flow Cytometry applications (Yun, T.J., et. al. (2016). Cell Metab. 23(5):852-66).
Research Category
Inflammation & Immunology

Quality

Evaluated by Immunohistochemistry in frozen mouse spleen tissue sections.

Immunohistochemistry Analysis: A 1:25 dilution of this antibody detected Siglec-H in frozen mouse spleen tissue sections.

Target description

34.35 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service