Skip to Content
Merck
All Photos(1)

Key Documents

MABF1976M

Sigma-Aldrich

Anti-Complement Receptor C5aR Antibody, clone 20/70

clone 20/70, from rat

Synonym(s):

C5a anaphylatoxin chemotactic receptor 1, C5a anaphylatoxin chemotactic receptor, C5a-R, C5aR, CD88

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€116.00
100 μG
€293.00

€116.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€116.00
100 μG
€293.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€116.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

20/70, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

activity assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
neutralization: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... C5Ar1(12273)

General description

C5a anaphylatoxin chemotactic receptor 1 (UniProt: P30993; also known as C5a anaphylatoxin chemotactic receptor, C5a-R, C5aR, CD88) is encoded by the C5ar1 (also known as C5ar, C5r1) gene (Gene ID: 12273) in murine species. C5aR is a multi-pass membrane protein of the G-protein coupled receptor 1 family that serves as a receptor for the chemotactic and inflammatory peptide anaphylatoxin C5a. The ligand interacts with at least two sites on the receptor: a high-affinity site on the extracellular N-terminus, and a second site in the transmembrane region which activates downstream signaling events. Receptor activation stimulates chemotaxis, granule enzyme release, intracellular calcium release, and superoxide anion production. C5aR is a homodimer, but can also form higher-order oligomers. C5aR becomes phosphorylated on serine residues in response to C5a binding that results in internalization of the receptor and short-term desensitization to C5a. The phosphorylated receptor is known to Interact with arrestin beta 1 and 2 (ARRB1 and 2) and this interaction is associated with internalization of C5aR. Sulfation is also reported to play a critical role in the association of C5aR with C5a, but it does not alter the ability of the receptor to transduce a signal and mobilize calcium in response to a small peptide agonist.

Specificity

Clone 20/70 detects complement 5a receptor in murine species. It targets an epitope with in the extracellular domain.

Immunogen

Rat basophilic leukemia RBL-2H3 cells stably expressing murine C5aR.

Application

Anti-Complement Receptor C5aR, clone 20/70, Cat. No. MABF1976, is a rat monoclonal antibody that detects Complement 5a receptor (C5aR) and has been tested for use in Flow Cytometry, Activity Assay, Inhibition of Activity/Function, Immunocytochemistry, and Neutralizing.
Neutralizing Analysis: A representative lot neutralized Complement receptor C5aR (Godau, J., et. al. (2004). J Immunol. 173(5):3437-45).

Immunocytochemistry Analysis: A representative lot detected Complement receptor C5aR in Immunocytochemistry applications (Soruri, A., et. al. (2003). Immunol Lett. 88(1):47-52).

Inhibits Activity/Function Analysis: A representative lot inhibited Complement receptor C5aR in Activity/Function applications (Godau, J., et. al. (2004). J Immunol. 173(5):3437-45; Soruri, A., et. al. (2003).

Flow Cytometry Analysis: A representative lot detected Complement receptor C5aR in Flow Cytometry applications (Shushakova, N., et. al. (2002). J Clin Invest. 110(12):1823-30).

Activity Assay Analysis: A representative lot detected Complement receptor C5aR in Activity Assay applications (Shushakova, N., et. al. (2002). J Clin Invest. 110(12):1823-30).

Quality

Evaluated by Flow Cytometry in BALB/c mouse bone marrow cells.

Flow Cytometry Analysis: 1 µg of this antibody detected Complement receptor C5aR in 1 million BALB/c mouse bone marrow cells.


Target description

39.02 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service