Skip to Content
Merck
All Photos(1)

Key Documents

MABF1966

Sigma-Aldrich

Anti-TSLP Receptor Antibody, clone 22H9

clone 22H9, from rat

Synonym(s):

Cytokine receptor-like factor 2, Cytokine receptor-like molecule 2, CRLM-2, Thymic stromal lymphopoietin protein receptor, Type I cytokine receptor delta 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€116.00
100 μG
€315.00

€116.00


Estimated to ship on03 April 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€116.00
100 μG
€315.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€116.00


Estimated to ship on03 April 2025


Request a Bulk Order

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

22H9, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Crlf2(57914)

General description

Cytokine receptor-like factor 2 (UniProt: Q8CII9; also known as Cytokine receptor-like molecule 2, CRLM-2, Thymic stromal lymphopoietin protein receptor, TSLP receptor, Type I cytokine receptor delta 1) is encoded by the Crlf2 (also known as rlm2, Tpte2, Tslpr) gene (Gene ID: 57914) in murine species. TSLP receptor is a single-pass type I membrane protein that serves as a receptor for thymic stromal lymphopoietin. It is expressed in liver, lung, testis, heart, brain, spleen, thymus, and bone marrow. Higher expression levels are observed in progenitors and myeloid cells. Its levels can be up-regulated in the myeloid 32D cell line by granulocyte colony-stimulating factor. TSLP receptor forms a functional complex with TSLP and IL7R, which is capable of stimulating cell proliferation through activation of STAT3 and STAT5. It has also been implicated in the development of the hematopoietic system. It is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off to generate the mature form. The mature form contains an extracellular domain (aa 20-232), a transmembrane domain (aa 233-253), and a cytoplasmic domain (aa 254-359). It contains WSXWS motif (aa 201-205), which is essential for proper protein folding and efficient intracellular transport and cell-surface binding. It also has a Box 1 motif (aa 262-270) that is required for JAK interaction and activation.

Specificity

Clone 22H9 is a rat monoclonal antibody that detects Thymic stromal lymphopoietin protein receptor (TSLPR) in murine species. It targets an epitope with in the extracellular domain.

Immunogen

Epitope: extracellular domain
His-tagged murine recombinant TSLP Receptor Protein.

Application

Anti-TSLP Receptor, clone 22H9, Cat. No. MABF1966, is a rat monoclonal antibody that detects TSLP receptor and has been tested for use in Flow Cytometry.
Flow Cytometry Analysis: A representative lot detected TSLP receptor in Flow Cytometry applications (Bell, B.D., et. al. (2013). Nat Immunol. 14(4):364-71; Isaksen, D.E., et. al. (2002). J Immunol. 168(7):3288-94; Iseki, M., et. al. (2012). Int Immunol. 24(3):183-95; Kashiwagi, M., et. al. (2017). Nat Immunol. 18(3):334-343).
Research Category
Inflammation & Immunology

Quality

Evaluated by Flow Cytometry in bone marrow cells from C57BL/6 Mouse.

Flow Cytometry Analysis: 1 µg of this antibody detected TSLP receptor in one million bone marrow cells from C57BL/6 Mouse.

Target description

37.76 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service