Skip to Content
Merck
All Photos(1)

Key Documents

MABF170D

Sigma-Aldrich

Anti-CD5 (human), PerCP, clone L17F12 Antibody

clone L17F12, from mouse, Peridinin-Chlorophyll-Protein Complex

Synonym(s):

T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, CD antigen CD5, CD5

Sign Into View Organizational & Contract Pricing

Select a Size

25 TESTS
€198.00
100 TESTS
€408.00
500 TESTS
€1,860.00

€198.00


Estimated to ship on07 May 2025


Request a Bulk Order

Select a Size

Change View
25 TESTS
€198.00
100 TESTS
€408.00
500 TESTS
€1,860.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€198.00


Estimated to ship on07 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

Peridinin-Chlorophyll-Protein Complex

antibody form

purified antibody

antibody product type

primary antibodies

clone

L17F12, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CD5(921)

Related Categories

General description

The L17F12 antibody is specific for human CD5, a 67 kD transmembrane glycoprotein that is expressed on most thymocytes, mature T cells, and a subset of B cells. CD5 is a member of the scavenger receptor superfamily and is present on approximately 70% of normal peripheral blood lymphocytes. CD5 is involved in modulating antigen receptor signaling in both T and B cells and acts through binding of CD72, its receptor expressed on B cells.

Application

Anti-CD5 (human), PerCP, clone L17F12 Antibody is an antibody against CD5 for use in Flow Cytometry.

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.06 μg from a representative lot detected CD5 in one million human peripheral blood lymphocytes.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service