Skip to Content
Merck
All Photos(1)

Key Documents

MABF119

Sigma-Aldrich

Anti-EGFR Antibody, clone 528 (Azide-free)

clone 528, from mouse

Synonym(s):

Epidermal growth factor receptor, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€446.00

€446.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€446.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

528, monoclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
neutralization: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... EGFR(1956)

General description

The epidermal growth factor receptor (EGFR; Proto-oncogene c-ErbB-1) is a ubiquitously receptor tyrosine kinase that is activated by a variety of ligands including EGF, EPGN, TGFA, and EREG. Ligand-bound EGFR undergoes dimerization and autophosphorylation exposing cytoplasmic domains which allow interaction with adaptor proteins and induction of several signaling pathways including PI3-AKT; PLCγ-PKC; Notch1; and Ras-Raf. EGFR therefore contributes to a diverse range of cellular processes inlcuding cell proliferation. Several studies have reported that EGFR signaling plays an important role in the development of various tumors. EGFR is regulated by PTPRJ and PTPRK phosphatases, and by the endocytic machinery including EPS15 that mediates internalization and degradation of the receptor.

Specificity

This antibody recognizes the extracellular domain of EGFR.

Immunogen

Partially purified human EGFR from A431 cells

Application

Anti-EGFR Antibody, clone 528 (Azide-free) detects level of EGFR & has been published & validated for use in Immunoprecipitation, Neutralizing.
Neutralizing Assay Analysis: A representative lot was used by an independent laboratory in A431 cells. (Kawamoto, T., et al. (1984). Journal of Biological Chemistry. 259(12):7761-7766.)

Quality

Evaluated by Immunoprecipitation in A431 cell lysate.

Immunoprecipitation Analysis: 10 µg of this antibody immunoprecipitated EGFR from A431 cell lysate.

Target description

~170 kDa observed. Uniprot gives a calculated molecular weight of 135 but can be observed at ~170 kDa due to phosphorylation. (Konishi, A., et al. (2003). The Journal of Biological Chemistry. 278(37):35049–35056.)

Linkage

Replaces: 04-338

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valencio Salema et al.
mAbs, 8(7), 1286-1301 (2016-07-30)
Most therapeutic antibodies (Abs) target cell surface proteins on tumor and immune cells. Cloning of Ab gene libraries in E. coli and their display on bacteriophages is commonly used to select novel therapeutic Abs binding target antigens, either purified or
Antoine Lesur et al.
Proteomics. Clinical applications, 9(7-8), 695-705 (2015-02-07)
We report an immunocapture strategy to extract proteins known to harbor driver mutations for a defined cancer type before the simultaneous assessment of their mutational status by MS. Such a method bypasses the sensitivity and selectivity issues encountered during the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service