Skip to Content
Merck

Skip To

MABE960

Anti-Fox1 Antibody, clone A2BP1

clone A2BP1, from rat

Synonym(s):

RNA binding protein fox-1 homolog 1, Ataxin-2-binding protein 1, Fox-1 homolog A, Hexaribonucleotide-binding protein 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

eCl@ss:
32160702
UNSPSC Code:
12352203
NACRES:
NA.46
Conjugate:
unconjugated
Clone:
A2BP1, monoclonal
Application:
ICC, IF, IHC (p)
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

A2BP1, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable, immunofluorescence: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RBFOX1(54715)
mouse ... Rbfox3(52897)

General description

42.78/44.79/40.01/42.08/42.40 kDa (human isoform 1/2 (gamma)/3/4 (alpha)/5 (beta)) and 42.68/40.49/44.73/40.71/40.29/42.34/38.92 kDa (mouse isoform 1 (A016)/2/3 (A713)/4/5 (A030)/6 (A715)/7 (A704)) calculated.
RNA binding protein fox-1 homolog 1 (UniProt Q9NWB1; also known as Ataxin-2-binding protein 1, Fox-1 homolog A, Hexaribonucleotide-binding protein 1) is encoded by the RBFOX1 (also known as A2BP, A2BP1, FOX1, HRNBP1) gene (Gene ID 54715) in human. The RNA-binding Fox (Rbfox) family of splicing factors is comprised of three members, Rbfox1 (Fox-1 or A2BP1), Rbfox2 (Fox-2 or RBM9), and Rbfox3 (Fox-3, HRNBP3 or NeuN). Rbfox proteins regulate splicing of many neuronal transcripts (pre-mRNAs) by binding the sequence (U)GCAUG in introns flanking alternative exons. A (U)GCAUG motif that lies downstream of the alternative exon generally promotes Rbfox-dependent exon inclusion, whereas an upstream motif will usually repress exon inclusion. Rbfox1 transcript itself is subjected to alternative splicing in response to chronic cell depolarization, resulting in the increased expression of the nuclear, splicing-active Rbfox1 isoform, which in turn leading to increased splicing of Rbfox1 target transcripts of proteins, including N-methyl D-aspartate (NMDA) receptor 1 (Grin1) and a calcium ATPase (Atp2b1), that play important roles in neuronal excitation and calcium homeostasis.

Immunogen

Recombinant full-length human Fox1.

Application

Anti-Fox1, clone A2BP1, Cat. No. MABE960, is a highly specific rat monoclonal antibody that targets Fox1 and has been tested in Immunocytochemistry, Immunofluorescence, and Immunohistochemistry.
Immunohistochemistry Analysis: A 1:50-1,000 dilution from a representative lot detected Fox1 in mouse cerebral cortex and heart tissue sections.

Immunocytochemistry Analysis: A representative lot immunostained exclusively differentiated neurons, but not neural stem cells, neuronal progenitors, astrocytes, or oligodendrocytes derived from p53-/-Pten+/- murine premalignant neural/progenitor stem cells (M-NSCs) cultured under various differentiation conditions (Hu, J., et al. (2013). Proc. Natl. Acad. Sci. U.S.A. 110(36):14520-14527).

Immunofluorescence Analysis: A representative lot immunostained postmitotic terminally differentiated neurons in brain sections from 2-week old postnatal mice (Hu, J., et al. (2013). Proc. Natl. Acad. Sci. U.S.A. 110(36):14520-14527).

Immunohistochemistry Analysis: A representative lot detected downregulated Fox1 immunoreactivity in more than 90% of human GBM tissue samples when compared with normal human brain tissues (Hu, J., et al. (2013). Proc. Natl. Acad. Sci. U.S.A. 110(36):14520-14527).
Research Category
Epigenetics & Nuclear Function

Biochem/physiol Actions

This monoclonal antibody immunostained exclusively differentiated neurons, but not neural stem cells, neuronal progenitors, astrocytes, or oligodendrocytes derived from p53-/-Pten+/- murine premalignant neural/progenitor stem cell (M-NSC) cultures (Hu, J., et al. (2013). Proc. Natl. Acad. Sci. U.S.A. 110(36):14520-14527).

Physical form

Format: Purified
Protein G purified.
Purified rat IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Immunohistochemistry in human cerebellum tissue section.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected Fox1 in human cerebellum tissue section.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters