Skip to Content
Merck

Skip To

MABE902

Anti-Pax-2 Antibody, clone 4C2.2

culture supernatant, clone 4C2.2, from mouse

Synonym(s):

Paired box protein Pax-2, Pax-2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
4C2.2, monoclonal
Application:
WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

4C2.2, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PAX2(5076)

General description

Paired box protein Pax-2 (UniProt Q02962; also known as Paired box homeotic gene 2) is encoded by the PAX2 (also known as FSGS7, PAPRS) gene (Gene ID 5076) in human. Pax-2 belongs to the family of transcription factors with conserved DNA-binding paired box domain. The family of paired box (PAX) genes is divided into four subgroups (I–IV). Genes in subgroups II (PAX2, PAX5, and PAX8) and III (PAX3 and PAX7) have been implicated in tumor development. PAX2 is a critical regulator of embryogenesis and it is normally expressed in the pancreas and in subpopulations of nodal lymphocytes in adult tissues. Aberrant PAX2 expression is frequently observed in several cancer types.
~45 kDa observed. ~45 kDa observed. Isoform 1 (44.7 kDa calculated); Isoform 2 (44.5 kDa calculated). An uncharacterized band ~17 kDa is seen in some lysate samples.

Immunogen

GST-tagged recombinant protein corresponding to human Pax-2.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-Pax-2 Antibody, clone 4C2.2 is validated for use in Western Blotting for the detection of Pax-2 .
Western Blotting Analysis: A 1:500 dilution from a representative lot detected PAX-2 in 10 µg of HeLa and PC3 cell lysate.

Biochem/physiol Actions

Clone 4C2.2 is expected to react with spliced Pax-2 isoform 1 and isoform 2 (Fetal kidney isoform). Reactivity toward isoforms 3&4 has not been tested.

Physical form

Mouse monoclonal IgG2bκ antibody in buffer containing supernatant without preservatives.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected PAX-2 in 10 µg of A431 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
WH0005076M1SAB2108774SAB2108773
species reactivity

human

species reactivity

human

species reactivity

-

species reactivity

-

antibody form

culture supernatant

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

mouse

biological source

mouse

biological source

rabbit

biological source

rabbit

Gene Information

human ... PAX2(5076)

Gene Information

human ... PAX2(5076)

Gene Information

human ... PAX2(5076)

Gene Information

human ... PAX2(5076)

conjugate

unconjugated

conjugate

unconjugated

conjugate

peroxidase conjugate

conjugate

FITC conjugate

clone

4C2.2, monoclonal

clone

3C7, monoclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters