Skip to Content
Merck

MABE897

Anti-ATRX Antibody

mouse monoclonal, 23C

Synonym(s):

ATRX, Transcriptional regulator ATRX, ATP-dependent helicase ATRX, X-linked helicase II, X-linked nuclear protein, XNP, Znf-HX

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Clone:
23C, monoclonal
Species reactivity:
mouse, human
Application:
ICC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-ATRX Antibody, clone 23C, clone 23C, from mouse

biological source

mouse

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

23C, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ATRX(546)

General description

Transcriptional regulator ATRX (EC 3.6.4.12; UniProt P46100; also known as Alpha thalassemia/mental retardation syndrome X-linked, ATP-dependent helicase ATRX, DNA dependent ATPase and helicase, Helicase 2 X-linked, X-linked helicase II, X-linked nuclear protein, XNP, Zinc finger helicase, Znf-HX) is encoded by the PRF1 (Also known as ATR2, JMS, MRXHF1, RAD54, RAD54L, SFM1, SHS, XH2, XNP, ZNF-HX) gene in human (Gene ID 546). Rad54 belongs to the SWI/SNF family of chromatin remodeling proteins. Rad54 undergoes cell cycle-dependent phosphorylation, nuclear matrix and chromatin association, and is implicatetd in playing a role in gene regulation during interphase and mitosis. ATRX mutations are associated with an X-linked mental retardation (XLMR) syndrome. most often accompanied by alpha-thalassemia syndrome, as a result of diverse changes in DNA methylation.
~283 kDa observed

Immunogen

Epitope: N-terminus
Recombinant protein corresponding to the N-terminus of human ATRX.

Application

Anti-ATRX Antibody, clone 23C is an antibody against ATRX for use in Western Blotting and Immunocytochemistry.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
Western Blotting Analysis: A representative lot detected ATRX in nuclear extracts from EBV-transformed lymphocytes (McDowell, T.L., et al. (1999). PNAS. 96(24):13983-13988).
Immunocytochemistry Analysis: A representative lot detected nuclear localization of ATRX in HeLa and L929 cells (McDowell, T.L., et al. (1999). PNAS. 96(24):13983-13988).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected ATRX in 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Explore all of our products under Anti-ATRX Antibody, clone 23C

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library