Skip to Content
Merck
All Photos(1)

Key Documents

MABE866

Sigma-Aldrich

Anti-Mad2 Antibody, clone AS55-A12

clone AS55-A12, from mouse

Synonym(s):

Mitotic spindle assembly checkpoint protein MAD2A, HsMAD2, Mitotic arrest deficient 2-like protein 1, MAD2-like protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€395.00

€395.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€395.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€395.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AS55-A12, monoclonal

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MAD2L1(4085)

General description

Mad2, also known as MAD2, MAD2L1, or Mitotic spindle assembly checkpoint protein MAD2A, is a mitotic spindle assembly checkpoint protein. The spindle checkpoint makes sure that all the chromosomes are aligned at the metaphase plate before anaphase begins which means that all chromosomes are attached to the spindle before anaphase can start. The spindle checkpoint becomes triggered when the chromosome(s) are unaligned. Mad2 then binds to the anaphase promoting complex (APC) and CDC20. This complex prevents the cell from transitioning into anaphase and ultimately from cell death or disease.

Immunogen

His-tagged recombinant protein corresponding to human Mad2.

Application

Anti-Mad2 Antibody, clone AS55-A12 is a highly specific mouse monoclonal antibody, that targets MAD2 & has been tested in western blotting & Immunofluorescence.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Western Blotting Analysis: A representative lot from an independent laboratory detected Mad2 in HeLa cell lysate (Santaguida, S., et al. (2011). EMBO J. 30(8):1508-1519.).

Immunofluorescence Analysis: A representative lot from an independent laboratory detected Mad2 in PtK1 cells transfected with Alexa-Mad2 (De Antoni, A., et al. 2005. Curr Biol. 15(3):214-225.).

Western Blotting Analysis: A representative lot from an independent laboratory detected Mad2 in HeLa cells transfected with pCMV expressing wild-type myc-tagged Mad2, and certain mutations of myc-tagged Mad2 (This mutant does not bind to the core of Mad1/Mad2) (De Antoni, A., et al. 2005. Curr Biol. 15(3):214-225.).

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Mad2 in 10 µg of HEK293 cell lyate.

Target description

~23 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katie L Dale et al.
Journal of cell science, 135(17) (2022-08-06)
Chromosomal instability (CIN), the process of increased chromosomal alterations, compromises genomic integrity and has profound consequences on human health. Yet, our understanding of the molecular and mechanistic basis of CIN initiation remains limited. We developed a high-throughput, single-cell, image-based pipeline
Timothy B Branigan et al.
Cell reports, 34(9), 108808-108808 (2021-03-04)
To identify genes whose loss confers resistance to CHK1 inhibitors, we perform genome-wide CRISPR-Cas9 screens in non-small-cell lung cancer (NSCLC) cell lines treated with the CHK1 inhibitor prexasertib (CHK1i). Five of the top six hits of the screens, MYBL2 (B-MYB)
Thomas Wild et al.
Cell reports, 25(9), 2317-2328 (2018-11-30)
The multisubunit ubiquitin ligase APC/C (anaphase-promoting complex/cyclosome) is essential for mitosis by promoting timely degradation of cyclin B1. APC/C is tightly regulated by the spindle assembly checkpoint (SAC), which involves MPS1 and MAD2-dependent temporal inhibition of APC/C. We analyzed the
Babhrubahan Roy et al.
Current biology : CB, 32(1), 237-247 (2021-12-04)
Accurate chromosome segregation during cell division requires amphitelic chromosome attachment to the spindle apparatus. It is ensured by the combined activity of the spindle assembly checkpoint (SAC),1 a signaling mechanism that delays anaphase onset in response to unattached chromosomes, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service