Skip to Content
Merck
All Photos(4)

Documents

MABE670

Sigma-Aldrich

Anti-Acetyl Histone H3 (Lys27) Antibody, clone 5E2.2

clone 5E2.2, from mouse

Synonym(s):

H3K27Ac, Histone H3 (acetyl K27), H3 histone family member T, histone cluster 3, H3t

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5E2.2, monoclonal

species reactivity

human

technique(s)

dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

acetylation (Lys27)

Gene Information

human ... H3C1(8350)

General description

Histone H3.1t (UniProt: Q16695; also known as H3K27Ac, Histone H3 (acetyl K27), H3 histone family member T, histone cluster 3, H3t) is encoded by the HIST3H3 (also known as H3FT) gene (Gene ID: 8290) in human. Histones are dynamic proteins that undergo multiple types of post-translational modifications, including acetylation, phosphorylation, methylation, and ubiquitination. Histone acetylation and deacetylation are reversible and play a major role in determining chromatin structure and function. Histone acetylation occurs mainly on the amino-terminal tail domains of histones. Acetylation neutralizes positive charges on histone and promotes the less condensed and more accessible chromatin conformation known as euchromatin. Histone acetylation is vital in regulating transcriptional activation and DNA replication, recombination, and repair. Histone H3 is one of the five ain histone proteins involved in the structure of chromatin in eukaryotic cells. It is the most extensively modified of the five histones. It features a main globular domain and a long N-terminal tail. Histone H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure.

Specificity

Clone 5E2.2 detects histone H3 acetylated on lysine27 in human.

Immunogen

KLH-cojugated peptide corresponding to a sequence around lysine 27 of yeast histone H3.

Application

Acetyl Histone H3 (Lys27) Antibody, clone 5E2.2, Cat. No. MABE670, is a highly specific mouse monoclonal antibody, that targets Acetyl Histone H3 (Lys27) and has been tested in Western Blotting and Immunocytochemistry and Dot Blot.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected Acetyl Histone H3 (Lys27) in Hela cells.



Dot Blot Analysis: A 1:1,000 dilution from a representative lot detected Acetyl Histone H3 (Lys27) in an Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and an Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665).


Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HeLa cells stimulated with Sodium Butyrate.


Western Blotting Analysis: 0.5 µg/mL of this antibody detected Acetyl Histone H3 (Lys27) in 10 µg of HeLa cells stimulated with Sodium Butyrate.

Target description

~17 kDa observed; 15.5 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: 05-1334

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suravi Pramanik et al.
Nucleic acids research, 50(6), 3394-3412 (2022-03-15)
Pancreatic ductal adenocarcinoma (PDAC), one of the most aggressive types of cancer, is characterized by aberrant activity of oncogenic KRAS. A nuclease-hypersensitive GC-rich region in KRAS promoter can fold into a four-stranded DNA secondary structure called G-quadruplex (G4), known to
Xiangxiang Wei et al.
Science advances, 5(3), eaau7887-eaau7887 (2019-03-21)
The transcription factor BTB and CNC homology 1 (Bach1) is expressed in the embryos of mice, but whether Bach1 regulates the self-renewal and early differentiation of human embryonic stem cells (hESCs) is unknown. We report that the deubiquitinase ubiquitin-specific processing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service