Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Clone:
RM190, monoclonal
Species reactivity:
human, rabbit
Application:
WB, multiplexing
Citations:
-
Pricing and availability is not currently available.
Product Name
Anti-Histone H3.3 Antibody, clone RM190,
antibody form
purified antibody
Quality Segment
clone
RM190, monoclonal
purified by
using Protein A
species reactivity
human, rabbit
concentration
(Please refer to lot specific datasheet.)
technique(s)
multiplexing: suitable, western blot: suitable
isotype
IgG
UniProt accession no.
General description
A peptide corresponding to human Histone H3.3.
~17 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Application
Multiplex Analysis (Mplex): 0.05-0.5 μg/mL from a representative lot was used in Multiplex.
Biochem/physiol Actions
This antibody reacts to Histone H3.3, independent of post-translational modifications. No cross reactivity with Histone H3.1 or other histone proteins.
Physical form
Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.
Analysis Note
Evaluated by Western Blotting in HeLa whole cell lysate, recombinant Histone H3.3 and recombinant Histone H3.1.
Western Blotting Analysis (WB): 1 μg/mL of this antibody detected Histone H3.3 in HeLa whole cell lysate and recombinant Histone H3.3.
Western Blotting Analysis (WB): 1 μg/mL of this antibody detected Histone H3.3 in HeLa whole cell lysate and recombinant Histone H3.3.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Explore all of our products under Anti-Histone H3.3 Antibody, clone RM190
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.