Skip to Content
Merck
All Photos(1)

Key Documents

MABE450

Sigma-Aldrich

Anti-TAF15, clone 8TA-2B10 Antibody

ascites fluid, clone 8TA-2B10, from mouse

Synonym(s):

TATA-binding protein-associated factor 2N, 68 kDa TATA-binding protein-associated factor, TAF(II)68, TAFII68, RNA-binding protein 56

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€432.00

€432.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€432.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€432.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

8TA-2B10, monoclonal

species reactivity

rat, mouse, human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TAF15(8148)

General description

TAF15, also known as TATA-binding protein-associated factor 2N, or 68 kDa TATA-binding protein-associated factor, or TAF(II)68, TAFII68, or RNA-binding protein 56, and encoded by the gene TAF15/RBP56/TAF2N, is a RNA and ssDNA binding protein important in transcription initiation. TAF15 aligns with the RNA polymerase II (Pol II) multiprotein complex that together with other proteins like TBP initiates transcription. TAF15 is ubiquitously expressed, and TAF15 shuttles from the nucleus to the cytoplasm upon activation; methylation of TAF15 appears to guide nuclear localization and transcriptional activity. Translocations involving the TAF15 gene may play a role in acute leukemia and extraskeletal myxoid chondrosarcoma, and mutations in this gene are associated with amyotrophic lateral sclerosis (ALS) diseases.

Application

Anti-TAF15 antibody, clone 8TA-2B10 is an antibody against TAF15 for use in western blotting & IP.

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected TAF15 in 10 µg of A431 cell lysate.

Target description

~68 kDa observed

Physical form

Mouse monoclonal IgG1κ ascites without preservatives.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Wang et al.
Oncotarget, 8(67), 111866-111881 (2018-01-18)
S-adenosyl methionine (SAM) is a ubiquitous methyl donor that was reported to have chemo- protective activity against liver cancer, however the molecular footprint of SAM is unknown. We show here that SAM selectively inhibits growth, transformation and invasiveness of hepatocellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service