Skip to Content
Merck
All Photos(2)

Key Documents

MABE337

Sigma-Aldrich

Anti-Hexim 1 Antibody, clone 10D2.1

clone 10D2.1, from mouse

Synonym(s):

Protein HEXIM1, Cardiac lineage protein 1, Estrogen down-regulated gene 1 protein, Hexamethylene bis-acetamide-inducible protein 1, Menage a quatre protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
€278.00

€278.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
200 μG
€278.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€278.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10D2.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HEXIM1(10614)

General description

Hexim1 is a transcriptional regulator which functions as a general RNA polymerase II transcription inhibitor. Hexim1 works in cooperation with 7SK snRNA to sequester P-TEFb in a large inactive 7SK snRNP complex, preventing RNA polymerase II phosphorylation and subsequent transcriptional elongation. Hexim1 may also regulate NF-kappa-B, ESR1, NR3C1 and CIITA-dependent transcriptional activity. The 7SK snRNP complex is composed of P-TEFb (composed of CDK9 and CCNT1/cyclin-T1), Hexim1, Hexim2, BCDIN3, SART3 proteins and 7SK and U6 snRNAs. Hexim1 also interacts with the N-CoR complex through NCOR1, ESR1, NR3C1, and may interact with NF-kappa-B through REL A. Hexim1 is located in the nucleus where it can bind with alpha-importin, and can also be found in the cytoplasm. Hexim1 is up-regulated by HMBA (hexamethylene bisacetamide) at the protein level and down-regulated by estrogen.

Immunogen

Epitope: C-terminus
GST-tagged recombinant protein corresponding to the C-terminus of human Hexim 1.

Application

Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected Hexim 1 in human placenta tissue.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-Hexim 1 Antibody, clone 10D2.1 is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of Hexim 1.

Quality

Evaluated by Western Blotting in human placenta tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Hexim 1 in 10 µg of human placenta tissue lysate.

Target description

~60 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service